Skip to Content
Merck
All Photos(8)

Key Documents

HPA017936

Sigma-Aldrich

Anti-YARS antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TyrRS, Anti-Tyrosyl-tRNA ligase, Anti-Tyrosyl-tRNA synthetase, cytoplasmic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SKEYTLDVYRLSSVVTQHDSKKAGAEVVKQVEHPLLSGLLYPGLQALDEEYLKVDAQFGGIDQRKIFTFAEKYLPALGYSKRVHLMNPMVPGLTGSKMSSSEEESKIDLLDRKEDVKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YARS(8565)

General description

Tyrosyl-tRNA synthetase (YARS) belongs to class I of the tRNA synthetase family. It has an amino-terminal catalytic core domain and an endothelial monocyte-activating polypeptide-II (EMAP II)-like carboxyl-terminal domain.

Immunogen

Tyrosyl-tRNA synthetase, cytoplasmic recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tyrosyl-tRNA synthetase (YARS) takes part in the two-step aminoacylation reaction, forming a tRNA bound to a tyrosyl moiety (Tyr-tRNATyr). YARS also takes part in DNA damage protection. During oxidative stress, it translocates from the cytosol to the nucleus and stimulates transcription factors like E2F1 to upregulate the transcription of genes involved in DNA damage repair. Mutations in the gene encoding YARS are associated with Charcot-Marie-Tooth disease (CMT). Studies have shown that it might be an important therapeutic target for chemotherapy-induced thrombocytopenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74193

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleksandr V Savytskyi et al.
Journal of molecular recognition : JMR, 26(2), 113-120 (2013-01-22)
Human tyrosyl-tRNA synthetase (HsTyrRS) is composed of two structural modules: N-terminal catalytic core and an EMAP II-like C-terminal domain. The structures of these modules are known, but no crystal structure of the full-length HsTyrRS is currently available. An all-atom model
Zhan Ling et al.
European journal of pharmacology, 738, 293-300 (2014-06-08)
Chemo- and radiotherapy induced thrombocytopenia present significant limitations for tumor therapy. In this study, we examined the therapeutic effect of recombinant human tyrosyl-tRNA synthetase (rhTyrRS) against development of thrombocytopenia in cyclophosphamide (CTX) treated mice. Our data indicate that intraperitoneal administration
Guangsen Fu et al.
The Journal of biological chemistry, 287(12), 9330-9334 (2012-02-01)
Aminoacyl-tRNA synthetases, essential components of the cytoplasmic translation apparatus, also have nuclear functions that continue to be elucidated. However, little is known about how the distribution between cytoplasmic and nuclear compartments is controlled. Using a combination of methods, here we
Na Wei et al.
Molecular cell, 56(2), 323-332 (2014-10-07)
Tyrosyl-tRNA synthetase (TyrRS) is known for its essential aminoacylation function in protein synthesis. Here we report a function for TyrRS in DNA damage protection. We found that oxidative stress, which often downregulates protein synthesis, induces TyrRS to rapidly translocate from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service