Skip to Content
Merck
All Photos(2)

Key Documents

HPA017851

Sigma-Aldrich

Anti-NDRG3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein NDRG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

MDELQDVQLTEIKPLLNDKNGTRNFQDFDCQEHDIETTHGVVHVTIRGLPKGNRPVIL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDRG3(57446)

General description

NDRG family member 3 (NDRG3) which is expressed in the testis and prostate gland, belongs to the N-myc down-regulated gene (NDRG) family. The gene encoding this protein is androgen regulated and located on chromosome 20q11.21-11.23. NDRG3 is a 363 amino acid polypeptide.

Immunogen

Protein NDRG3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NDRG family member 3 (NDRG3) maybe involved in spermatogenesis. It has been shown that its expression and upregulation contributes to the advancement of prostate cancer. This protein can be used as a diagnostic marker for hepatitis B virus-related hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74471

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Guang Fan et al.
Oncology reports, 26(5), 1281-1286 (2011-07-05)
microRNAs (miRNAs) are short, non-coding RNAs with post-transcriptional regulatory functions that participate in diverse biological pathways. miR-122, a liver-specific miRNA, has been found to be down-regulated in hepatocellular carcinoma (HCC) and HCC-derived cell lines. In this study, miR-122 was down-regulated
Guo-feng Ren et al.
Histology and histopathology, 29(4), 535-542 (2013-11-14)
To investigate the clinicopathologic significance of NDRG2 and NDRG3, and their involvement in recurrence-free survival (RFS) and overall survival (OS) of prostate cancer (PCa). NDRG2 and NDRG3 expression in 206 pairs of primary PCa and corresponding noncancerous prostate tissue samples
Weiqun Wang et al.
International journal of cancer, 124(3), 521-530 (2008-11-01)
NDRG3 belongs to the N-myc down-regulated gene (NDRG) family that contains 4 paralogs: NDRG1, -2, -3 and -4. The function of NDRG3 and its relationship to cancer has not been studied. We herein report our examination of the expression and
W Zhao et al.
Biochimica et biophysica acta, 1519(1-2), 134-138 (2001-06-19)
We report the cloning and expression pattern of a novel N-myc downstream-regulated gene 3 (NDRG3), located on human chromosome 20q11.21-11.23. The NDRG3 cDNA is 2588 base pair in length, encoding a 363 amino acid polypeptide highly related to mouse Ndr3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service