Skip to Content
Merck
All Photos(4)

Key Documents

HPA017055

Sigma-Aldrich

Anti-SAMSN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hematopoietic adaptor containing SH3 and SAM domains 1, Anti-SAM domain, SH3 domain and nuclear localization signals protein 1, Anti-SAM domain-containing protein SAMSN-1, Anti-SH3-SAM adaptor protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LLNGYETLEDLKDIKESHLIELNIENPDDRRRLLSAAENFLEEEIIQEQENEPEPLSLSSDISLNKSQLDDCPRDSGCYISSGNSDNGKEDLESENLSDMVHKIIITEPS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SAMSN1(64092)

General description

SAMSN1 (SAM domain, SH3 domain and nuclear localization signals 1) is a cytoplasmic adaptor protein belonging to the SLY family. It is abundantly expressed in the hematopoietic compartment. Its expression to a lesser extent have also been found in heart, brain, placenta, and lung. Generally, they are characterized by the presence of protein-protein interaction domains. It consists of a sterile motif (SAM) domain that is capable of self-association and can also associate with each other. The chimeric fusion of this domain with the ßPDGF receptor, AML1, c-Abl, and JAK2 results in the oncogenic transformation of the SAM domain. The SH3 domains mediate protein-protein interactions in several signal transduction pathways.[1][2]

Immunogen

SAM domain-containing protein SAMSN-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SAMSN1 (SAM domain, SH3 domain and nuclear localization signals 1) protein may be involved in the formation of blood vessel during normal brain development. It may function as an adaptor or scaffold protein. It has been found to be overexpressed in glioma cells and is associated with a poor survival in glioblastoma multiforme. SAMSN1 may be involved in B cell activation and differentiation. SAMSN1 has been suggested to be a tumor suppressor in lung cancer.[1][2] It has also been reported that SAMSN1 may act as a biomarker for the phenotype of human hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71505

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression of SAMSN1 Expression is Associated with the Malignant Phenotype of Hepatocellular Carcinoma.
Satoshi S
Annals of Surgical Oncology, 1-8 (2015)
Jacqueline E Noll et al.
Neoplasia (New York, N.Y.), 16(7), 572-585 (2014-08-15)
Multiple myeloma (MM), a hematological malignancy characterized by the clonal growth of malignant plasma cells (PCs) in the bone marrow, is preceded by the benign asymptomatic condition, monoclonal gammopathy of undetermined significance (MGUS). Several genetic abnormalities have been identified as
Yong Yan et al.
PloS one, 8(11), e81905-e81905 (2013-11-28)
To study the expression pattern and prognostic significance of SAMSN1 in glioma. Affymetrix and Arrystar gene microarray data in the setting of glioma was analyzed to preliminarily study the expression pattern of SAMSN1 in glioma tissues, and Hieratical clustering of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service