Skip to Content
Merck
All Photos(6)

Key Documents

HPA016852

Sigma-Aldrich

Anti-PTDSS1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PSS-1, Anti-Phosphatidylserine synthase 1, Anti-PtdSer synthase 1, Anti-Serine-exchange enzyme I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AEHYGHREKTYSECEDGTYSPEISWHHRKGTKGSEDSPPKHAGNNESHSSRRRNRHSKSKVTNGVGKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTDSS1(9791)

Immunogen

Phosphatidylserine synthase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PTDSS1 (Phosphatidylserine synthase 1) is an integral membrane protein involved in the phosphatidylserine synthase I activity. It is abundantly expressed in all murine tissues of kidney, brain, and liver. It catalyzes the base-exchange reaction to convert PC (Phosphatidylcholine) and PE (phosphatidylethanolamine) respectively into PS (phosphatidylserine) in intact cells. Heterozygous missense mutations of PTDSS1 cause Lenz-Majewski syndrome (LMS) syndrome with intellectual disability and multiple congenital anomalies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74647

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sérgio B Sousa et al.
Nature genetics, 46(1), 70-76 (2013-11-19)
Lenz-Majewski syndrome (LMS) is a syndrome of intellectual disability and multiple congenital anomalies that features generalized craniotubular hyperostosis. By using whole-exome sequencing and selecting variants consistent with the predicted dominant de novo etiology of LMS, we identified causative heterozygous missense
B Sturbois-Balcerzak et al.
The Journal of biological chemistry, 276(11), 8205-8212 (2000-11-21)
In mammalian cells, phosphatidylserine is synthesized by two different enzymes, phosphatidylserine synthase (PSS)-1 and -2, via a base exchange reaction in which the head group of a phospholipid (phosphatidylcholine or phosphatidylethanolamine) is replaced by l-serine. Since the amino acid sequences
Shiho Tomohiro et al.
The Biochemical journal, 418(2), 421-429 (2008-11-19)
PS (phosphatidylserine) in mammalian cells is synthesized by two distinct base-exchange enzymes, PSS1 (PS synthase 1) and PSS2, which are responsible for the conversion of PC (phosphatidylcholine) and PE (phosphatidylethanolamine) respectively into PS in intact cells. The PS synthesis in
O Kuge et al.
The Journal of biological chemistry, 266(35), 24184-24189 (1991-12-15)
A phosphatidylserine-auxotrophic mutant of cultured Chinese hamster ovary (CHO) cells, PSA-3, is defective in phosphatidylserine synthase I activity. Transfection of PSA-3 cells with a cDNA expression library of CHO-K1 (the parent of PSA-3) yielded a phosphatidylserine-prototrophic transformant with normal phosphatidylserine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service