Skip to Content
Merck
All Photos(4)

Key Documents

HPA016812

Sigma-Aldrich

Anti-RNF41 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NRDP1, Anti-SBBI03

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

DAVLQAVIKRSLVESGCPASIVNELIENAHERSWPQGLATLETRQMNRRYYENYVAKRIPGKQAVVVMACENQHMGDDMVQEPGLVMIFAHGVEEI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF41(10193)

General description

RNF41 (Ring finger protein 41) is an E3 ubiquitin ligase consisting of RING finger domain including ErbB3 and Parkin.

Immunogen

ring finger protein 41, E3 ubiquitin protein ligase

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RNF41 (Ring finger protein 41) is involved in controlling several protein activity during cell growth, apoptosis and oxidative stress. In addition, it plays a potential role in the tumor progression and pathogenesis of Parkinson′s disease. It acts as a pro-apoptotic protein in human glioma to slow down the cell apoptosis. It aggravates the rate of tumor progression by low apoptotic rate. It has been suggested that RNF41 may play an important regulatory role in the development of human glioma. It also has an impact on the RNF41-mediated ErbB3 degradation, which retards the cellular growth of colorectal cancer (CRC). Studies also have reported that reduced RNF41 expression in CRC may promote tumor progression, which indicates it being a novel target for CRC therapy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73761

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hang Lu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(9), 8639-8643 (2014-05-29)
The molecular mechanism underlying the proliferation of colorectal cancer (CRC) cells is not completely understood. Here, we found that the level of neuregulin receptor degradation protein-1 (Nrdp1) E3 ubiquitin ligase was significantly decreased in CRC tissues, compared with the adjacent
Hengliang Shi et al.
IUBMB life, 66(10), 704-710 (2014-10-31)
Ubiquitin ligase Nrdp1 (neuregulin receptor degradation protein 1) plays important roles in multiple physiological process because it can ubiquitinate various substrates such as ErbB3, BRUCE, MyD88, C/EBPβ, and Parkin, and so forth. In addition to the physiological function, it was
An Zhou et al.
Biochemical and biophysical research communications, 410(4), 771-774 (2011-06-23)
Nrdp1 is a RING finger containing ubiquitin E3 ligase that interacts with and modulates activity of multiple proteins, including ErbB3 and Parkin, a causative protein for early onset recessive juvenile parkinsonism (AR-JP). To investigate the functions of Nrdp1, we have
Kirsteen M Tullett et al.
eLife, 9 (2020-12-03)
The dendritic cell receptor Clec9A facilitates processing of dead cell-derived antigens for cross-presentation and the induction of effective CD8+ T cell immune responses. Here, we show that this process is regulated by E3 ubiquitin ligase RNF41 and define a new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service