Skip to Content
Merck
All Photos(1)

Key Documents

HPA016708

Sigma-Aldrich

Anti-PHF11 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BRCA1 C-terminus-associated protein, Anti-PHD finger protein 11, Anti-Renal carcinoma antigen NY-REN-34

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

DATVKVPFLKKCKEAGLLNYLLEEILDKVHSIPEKLMDETTSESDYEEIGSALFDCRLFEDTFVNFQAAIEKKIHASQQRWQQLKEEIELLQDLKQTLCSFQENRDLMSSSTSISSLSY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF11(51131)

Immunogen

PHD finger protein 11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PHF11 (PHD finger protein 11) is a plant homeodomain zinc finger protein, hypothesized to be involved in chromatin-mediated transcriptional regulation and immunoregulation, primarily in B-lymphocytes. It′s expression was proven to be higher in TH1-type cells, thereby making it regulator of TH1-type cytokine gene expression. Hence polymorhism in PHF11 can lead to reduction in its expression, increased serum IgE levels, therby leading to asthma and severe atopic dermatitis (AD) in childre..

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73713

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Jang et al.
Genes and immunity, 6(3), 262-264 (2005-01-28)
Atopic dermatitis is an increasingly common and debilitating childhood disorder that is often accompanied by asthma and allergic rhinitis. Although the pathology of these disorders is distinct, the majority of cases are atopic, typified by elevated serum IgE. A new
J McClenaghan et al.
Thorax, 64(7), 620-625 (2009-04-24)
Numerous areas of the human genome have previously been associated with asthma and asthma-related phenotypes, but few positive findings have been successfully replicated in independent populations. Initial studies have reported strong associations of variants in the plant homeodomain zinc finger
Emily Clarke et al.
The Journal of allergy and clinical immunology, 121(5), 1148-1154 (2008-04-15)
Polymorphisms in the plant homeodomain finger protein 11 gene (PHF11) are associated with increased total serum IgE levels, asthma, and severe atopic dermatitis (AD) in children. Although PHF11 includes a plant homeodomain, a motif often found in transcriptional regulators, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service