Skip to Content
Merck
All Photos(6)

Key Documents

HPA015662

Sigma-Aldrich

Anti-SEMA4D antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A8, Anti-BB18, Anti-CD100 antigen, Anti-GR3, Anti-Semaphorin-4D precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LLIGKKKPKSDFCDREQSLKETLVEPGSFSQQNGEHPKPALDTGYETEQDTITSKVPTDREDSQRIDDLSARDKPFDVKCELKFADSDADGD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEMA4D(10507)

Immunogen

Semaphorin-4D precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Semaphorin-4D (SEMA4D) is a protein is encoded by the SEMA4D gene in humans and belongs to semaphorin family. It functions in promoting tumor proliferation, migration and metastasis when overexpressed. It plays an important role in Akt protein phosphorylation and its inhibition may be helpful in treating NSCLC (Non-small-cell lung carcinoma) and cervical cancer. It may also play some role in promoting epithelial ovarian cancers (EOCs).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72991

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Chen et al.
Asian Pacific journal of cancer prevention : APJCP, 14(10), 5883-5890 (2013-12-03)
Previously, we demonstrated overexpression of semaphorin4D (SEMA4D, CD100) to be closely related to tumor angiogenesis in epithelial ovarian cancers (EOCs). However, the function and expression of SEMA4D in the EOC microenvironment has yet to be clarified in detail. In this
Sha-sha Ruan et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 34(4), 491-496 (2014-08-20)
This study aimed to explore Semaphrin4D (Sema4D) expression and clinical significance in non-small cell lung cancer (NSCLC), and to define the roles and mechanisms of Sema4D in regulating the malignant behaviors of A549 cells by small interfering RNA (siRNA). Firstly
Tetsuro Ikeya et al.
BMC cancer, 16, 525-525 (2016-07-28)
Binding to Sema4D and PlexinB1 induce angiogenesis and invasive growth in colorectal cancer (CRC). The expression of Semaphorin4D (Sema4D) and PlexinB1 has been shown to be related to the prognosis of patients with various malignancies. However, the correlation between the
Huidong Liu et al.
Microvascular research, 93, 1-8 (2014-03-08)
Lymphangiogenesis is thought to be essential for cancer progression, making it an important target in cancer therapy. Lymphangiogenic factors (VEGF-C and VEGF-D) are upregulated in various tumors/cancers, and play an important role in lymphangiogenesis and lymph node metastasis. Similarly, semaphorin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service