Skip to Content
Merck
All Photos(6)

Key Documents

HPA015325

Sigma-Aldrich

Anti-CDK17 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PCTAIRE- motif protein kinase 2, Anti-PCTK2 antibody produced in rabbit, Anti-Serine/threonine-protein kinase PCTAIRE-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RRLSLTLRGSQTIDESLSELAEQMTIEENSSKDNEPIVKNGRPPTSHSMHSFLHQYTGSFKKPPLRRPHSVIGGSLGSFMAMPRNGSRLDIVHENLKMGSDGESDQASGTSSDEVQSPTGVCLRNRI

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCTK2(5128)

General description

CDK17 (cyclin-dependent kinase 17) belongs to a subfamily of Cdc2-related kinases, and is exclusively expressed in brain. It is expressed usually in post-mitotic cells. In brains, it has predominant expression in olfactory bulb and hippocampus, which are generally made of post-mitotic neurons. This gene is localized to human chromosome 12q23.1, which codes for a protein composed of 523 amino acids, and has a molecular weight of 60kDa. It contains a central kinase domain and one N- and C-terminal domain each.

Immunogen

Serine/threonine-protein kinase PCTAIRE-2 recombinant protein epitope signature tag (PrEST)

Application

Anti-CDK17 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CDK17 (cyclin-dependent kinase 17) acts as a serine/threonine kinase, and phosphorylates histone H1. It is an interacting partner of Trap (tudor repeat associator with PCTAIRE 2), and it interacts with it through its N-terminal.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73465

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam R Cole
Neuro-Signals, 17(4), 288-297 (2009-10-10)
PCTAIRE kinases (PCTKs) are highly conserved serine/threonine kinases that are closely related to cyclin-dependent kinases. They are enriched in post-mitotic neurons of adult brains, suggesting they might perform important neuron-specific functions independent of the cell cycle. So far, the biological
T Hirose et al.
European journal of biochemistry, 267(7), 2113-2121 (2000-03-23)
PCTAIRE 2 is a Cdc2-related kinase that is predominantly expressed in the terminally differentiated neuron. To elucidate the function of PCTAIRE 2, proteins that associate with PCTAIRE 2 were screened by the yeast two-hybrid system. A positive clone was found
T Hirose et al.
European journal of biochemistry, 249(2), 481-488 (1997-11-25)
PCTAIRE are members of a subfamily of Cdc2-related kinases that have been shown to be preferentially expressed in post-mitotic cells. To examine the neural functions of PCTAIRE, rat cDNA clones encoding PCTAIRE 1, 2, and 3 were isolated, and their
Rochelle C J D'Souza et al.
Science signaling, 7(335), rs5-rs5 (2014-07-25)
Transforming growth factor-β (TGF-β) signaling promotes cell motility by inducing epithelial-to-mesenchymal transitions (EMTs) in normal physiology and development, as well as in pathological conditions, such as cancer. We performed a time-resolved analysis of the proteomic and phosphoproteomic changes of cultured
Time-resolved dissection of early phosphoproteome and ensuing proteome changes in response to TGF-?.
D'Souza RC, Knittle AM, Nagaraj N, et al.
Science Signaling, 7(335), rs5-rs5 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service