Skip to Content
Merck
All Photos(3)

Key Documents

HPA014957

Sigma-Aldrich

Anti-BRI3BP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BRI3-binding protein, Anti-Cervical cancer 1 proto- oncogene-binding protein KG19, Anti-HCCR-1, Anti-I3-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MFVETLWKVWTELLDVLGLDVSNLSQYFSPASVSSSPARALLLVGVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRI3BP(140707)

General description

BRI3BP (BRI3 binding protein) is an oncogene, which is a HCCR (human cervical cancer)-interacting protein, and is also called HCCRBP-1. This protein is composed of 166 amino acids, and has a molecular weight of 19kDa. It is also called BASH N-terminus-associated protein 1 (BNAS1), and contains four putative transmembrane regions and a leucine-zipper. It has a ubiquitous expression pattern. It resides in the endoplasmic reticulum.

Immunogen

BRI3-binding protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BRI3BP (BRI3 binding protein) is an oncogene, which is up-regulated in multiple tumors. It induces protein kinase C (PKC) activities, while suppressing the function of PKCδ and PKCα isoforms, which are pro-apoptotic in nature. It also stabilizes p53 gene, thus promoting tumorigenesis. It interacts with BASH/BLNK (B-cell linker)/SLP-65 protein, at the N-terminal, at the membrane of either endoplasmic reticulum (ER) or nucleus. It activates Elk-1, by regulation of the signaling pathway through mitogen-activated protein kinase (MAPK). It also affects the ER structural dyamics, as well as regulates mitochondrial viability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72345

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seon-Ah Ha et al.
International journal of cancer, 122(3), 501-508 (2007-10-19)
Oncogene HCCR-1 functions as a negative regulator of the p53 and contributes to tumorigenesis of various human tissues. HCCR transgenic mice developed breast cancers but it is unknown how HCCR-1 contributes to human tumorigenesis. This study identified a HCCR-1-binding protein
Takashi Katahira et al.
International immunology, 18(4), 545-553 (2006-02-17)
BASH/BLNK/SLP-65 is an adaptor protein necessary for the B cell receptor (BCR) signal transduction. Here we report the identification through the yeast two-hybrid system of a novel 26-kDa protein, BASH N-terminus-associated protein 1 (BNAS1), which interacts with the conserved and
Tetsuo Yamazaki et al.
Biochemical and biophysical research communications, 362(4), 971-975 (2007-09-04)
To determine the contribution of the endoplasmic reticulum (ER) to cell fate decision, we focused on BRI3-binding protein (BRI3BP) residing in this organelle. BRI3BP, when overexpressed, augmented the apoptosis of human embryonic kidney 293T cells challenged with drugs including the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service