Skip to Content
Merck
All Photos(2)

Key Documents

HPA014948

Sigma-Aldrich

Anti-SLC37A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Solute carrier family 37 member 2, Anti-Sugar phosphate exchanger 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RKPISIVKSRLHQNCSEQIKPINDTHSLNDTMWCSWAPFDKDNYKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

SLC37A2 (solute carrier family 37, member 2) is a phosphate (Pi)-associated antiporter, which resides in the endoplasmic reticulum (ER). It is a member of the solute carrier 37 (SLC37) family, which consists of four members. It is expressed in thymus, spleen, liver, kidney, pancreas, intestines and macrophages. This gene is localized to human chromosome 11q24.2, contains 18 coding exons, and codes for a protein with predicted 501 amino acids. It might be a member of the major facilitator superfamily (MFS) as it contains twelve transmembrane domains. It has three putative N-glycosylation sites.

Immunogen

Sugar phosphate exchanger 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC37A2 (solute carrier family 37, member 2) is a glucose-6-phosphate (G6P) antiporter. During differentiation of macrophages, its expression is elevated. This may show a link between differentiation and G6P and SLC37A2. Studies show that as a transporter, it has an essential role in macrophage metabolism, thus, also playing a role in immunity. Its expression is related to obese white adipose tissue.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73286

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Young Kim et al.
American journal of physiology. Endocrinology and metabolism, 293(1), E110-E120 (2007-03-16)
A marked degree of macrophage infiltration of white adipose tissue (WAT) occurs in obesity and may link excess adiposity with the chronic inflammatory state underlying metabolic syndrome and other comorbidities of obesity. Excess deposition of fat in the intra-abdominal vs.
Janice Y Chou et al.
Current topics in membranes, 73, 357-382 (2014-04-22)
The SLC37 family members are endoplasmic reticulum (ER)-associated sugar-phosphate/phosphate (P(i)) exchangers. Three of the four members, SLC37A1, SLC37A2, and SLC37A4, function as Pi-linked glucose-6-phosphate (G6P) antiporters catalyzing G6P:P(i) and P(i):P(i) exchanges. The activity of SLC37A3 is unknown. SLC37A4, better known
Chi-Jiunn Pan et al.
PloS one, 6(9), e23157-e23157 (2011-09-29)
Blood glucose homeostasis between meals depends upon production of glucose within the endoplasmic reticulum (ER) of the liver and kidney by hydrolysis of glucose-6-phosphate (G6P) into glucose and phosphate (P(i)). This reaction depends on coupling the G6P transporter (G6PT) with
Lucia Bartoloni et al.
Pflugers Archiv : European journal of physiology, 447(5), 780-783 (2003-06-18)
The SLC37 family of four predicted proteins is an almost unexplored group of transmembrane sugar transporters. Of the four proteins/genes assigned to date to this family, only one is well known, the SLC37A4 gene (also known as the glucose-6-phosphate transporter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service