Skip to Content
Merck
All Photos(2)

Key Documents

HPA014722

Sigma-Aldrich

Anti-KCNJ5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CIR, Anti-GIRK4, Anti-KATP1, Anti-Kir3.4, Anti-LQT13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

TPVLTLEKGFYEVDYNTFHDTYETNTPSCCAKELAEMKREGRLLQYLPSPPLLGGCAEAGLDAEAEQNEEDEPKGLGG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNJ5(3762)

General description

KCNJ5 (potassium channel, inwardly rectifying subfamily J, member 5) is an inward rectifier K+ channel 4, which is activated by G-protein. This protein is also called Kir3.4, and is expressed in zona glomerulosa, and forms either homotetramers or heterotetramers with Kir3.1 (KCNJ3), Kir3.2 (KCNJ6) or Kir3.3 (KCNJ9). The pore of this channel has a GYG motif at its narrowest position, which acts as a selectivity filter against K+. KCNJ5 contains four subunits, with each of them spanning the membrane twice. The pore is present in between the four subunits. This gene is localized to human chromosome 11q24.

Immunogen

G protein-activated inward rectifier potassium channel 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KCNJ5 (potassium channel, inwardly rectifying subfamily J, member 5) is a channel that acts as a filter to reduce the K+ selectivity of cells. This leads to the movement of Na+ ions inside the cell, causing depolarization, which eventually results in enhanced synthesis of aldosterone. Hyperpolarization of membrane by this channel leads to inhibition of prolactin secretion by pituitary lactotropes, when acted upon by dopamine. Mutations in this gene are associated with familial hyperaldosteronism type III (FH-III), and are also involved in the pathophysiology of sporadic aldosterone-producing adenomas. Polymorphisms in this gene are linked to common as well as rare forms of primary aldosteronism. Mutations in KCNJ5 might also be linked with unilateral adrenal hyperplasia. Polymorphisms in this gene are also associated with Tourette Syndrome (TS) and attention-deficit/hyperactivity disorder (ADHD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72857

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Gomez et al.
Genes, brain, and behavior, 13(6), 535-542 (2014-05-21)
Linkage and association of Tourette Syndrome (TS) and Attention-Deficit/Hyperactivity Disorder (ADHD) have previously been reported in the 11q24 chromosomal region. To identify the risk gene within the region we studied the potassium inwardly-rectifying channel J5 (KCNJ5) gene in a sample
Jingjing Zhang et al.
Zhonghua yi xue yi chuan xue za zhi = Zhonghua yixue yichuanxue zazhi = Chinese journal of medical genetics, 32(1), 21-25 (2015-01-31)
To investigate the prevalence of KCNJ5 gene missense mutations and their role in patients with unilateral adrenal hyperplasia (UAH). Fourteen UAH tissues were collected through surgical resection, and all the tissues were confirmed by pathology. Peripheral blood samples of the
Silvia Monticone et al.
The Journal of clinical endocrinology and metabolism, 98(11), E1861-E1865 (2013-09-17)
Primary aldosteronism is a heterogeneous group of disorders comprising both sporadic and familial forms. Mutations in the KCNJ5 gene, which encodes the inward rectifier K(+) channel 4 (G protein-activated inward rectifier K(+) channel 4, Kir3.4), cause familial hyperaldosteronism type III
Meena Murthy et al.
Hypertension (Dallas, Tex. : 1979), 63(4), 783-789 (2014-01-15)
Primary aldosteronism (autonomous aldosterone production with suppressed renin) plays an important pathophysiological role in what has been previously labeled as essential hypertension. Besides the recently described germline mutations in the KCNJ5 potassium channel associated with familial primary aldosteronism, somatic mutations
Carolina Velarde-Miranda et al.
Clinical and experimental pharmacology & physiology, 40(12), 895-901 (2013-07-09)
The G-protein-activated inwardly rectifying potassium channel Kir3.4 is expressed in the zona glomerulosa cell membrane and transports potassium out of the cell.  Angiotensin II stimulation of aldosterone secretion is mediated, in part, by suppression of the transcription of KCNJ5, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service