Skip to Content
Merck
All Photos(2)

Key Documents

HPA014288

Sigma-Aldrich

Anti-ARHGAP24 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FilGAP, Anti-Filamin-A-associated RhoGAP, Anti-RAC1- and CDC42-specific GTPase-activating protein of 72 kDa, Anti-RC-GAP72, Anti-Rho GTPase-activating protein 24, Anti-Rho-type GTPase-activating protein 24, Anti-RhoGAP of 73 kDa, Anti-Sarcoma antigen NY-SAR-88, Anti-p73RhoGAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ILNSDTLGNPTNVRNMSWLPNGYVTLRDNKQKEQAGELGQHNRLSTYDNVHQQFSMMNLDDKQSIDSATWSTSSCEISLPENSNSCRSSTTTCPEQDFFGGNFEDPVLDGPPQDDLSHPRDYESKSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The gene ARHGAP24 (Rho GTPase activating protein 24) encodes a member of the RhoGAP family proteins that are characterized by the presence of Pleckstrin homology (PH) domain and C-terminal coiled-coil domain.

Immunogen

Rho GTPase-activating protein 24 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Rho GTPase activating protein 24 encoded by the gene ARHGAP24 functions as a regulator of angiogenesis. It has Rho GTPase activity and plays a crucial role in endothelial cell functions, such as migration, proliferation, and capillary-tube formation. This vascular cell-specific GTPase-activating protein is found to be up-regulated in angiogenic milieu and may serve as a potential drug target for angiogenesis. The C-terminal region interacts with actin stress fibers, focal adhesions, and cell-cell adherens junctions, and regulates cell morphology and cytoskeletal organization. It also binds filamin A and controls actin remodeling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71496

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irena Lavelin et al.
The Journal of biological chemistry, 280(8), 7178-7185 (2004-12-22)
In the present study we characterize a novel RhoGAP protein (RC-GAP72) that interacts with actin stress fibers, focal adhesions, and cell-cell adherens junctions via its 185-amino acid C-terminal region. Overexpression of RC-GAP72 in fibroblasts induces cell rounding with partial or
Yasutaka Ohta et al.
Nature cell biology, 8(8), 803-814 (2006-07-25)
FilGAP is a newly recognized filamin A (FLNa)-binding RhoGTPase-activating protein. The GTPase-activating protein (GAP) activity of FilGAP is specific for Rac and FLNa binding targets FilGAP to sites of membrane protrusion, where it antagonizes Rac in vivo. Dominant-negative FilGAP constructs
Masuko Katoh et al.
International journal of molecular medicine, 14(2), 333-338 (2004-07-16)
RhoGAP family proteins, encoded by ARHGAP family genes, are negative regulators of Rho family GTPases, which are implicated in actin remodeling, cell polarity control, and cell migration. Based on the homology with ARHGAP22, we identified and characterized two novel ARHGAP
Zhi-Jian Su et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(33), 12212-12217 (2004-08-11)
Angiogenesis is a major therapeutic target. Ideal drug targets are genes expressed only in endothelial cells (ECs) or only during the angiogenic process. Here, we describe a gene, p73RhoGAP (p73), that has both of these properties. By using a PCR-based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service