Skip to Content
Merck
All Photos(4)

Key Documents

HPA013819

Sigma-Aldrich

Anti-ACKR2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-C chemokine receptor D6, Anti-CCBP2, Anti-Chemokine receptor CCR-10, Anti-Chemokine receptor CCR-9, Anti-Chemokine-binding protein 2, Anti-Chemokine-binding protein D6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QYLKAFLAAVLGWHLAPGTAQASLSSCSESSILTAQEEMTGMNDLGERQSENYPNKEDVGNKSA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCBP2(1238)

General description

The gene ACKR2 (atypical chemokine receptor 2) is mapped to human chromosome 3p21. It encodes a protein containing a conserved tyrosine motif at the N terminus that is involved in ligand binding, internalization, and scavenging. It is found to be expressed in barrier tissues, such as skin, gut, lung, and syncytiotrophoblast layer of the placenta. In adults, it is found to be expressed on lymphatic endothelial cells, leukocytes and keratinocytes.

Immunogen

Chemokine-binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene ACKR2 (atypical chemokine receptor 2) encodes a chemokine receptor that binds, internalizes, and degrades inflammatory CC-chemokines and regulates inflammatory responses that are stimulated by chemokines. It can bind up to 14 different inflammatory CC-chemokines. It facilitates the removal of chemokines from inflamed sites.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72668

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Eun Oh et al.
Nature, 571(7763), 122-126 (2019-06-14)
Antibodies secreted into mucosal barriers serve to protect the host from a variety of pathogens, and are the basis for successful vaccines1. In type I mucosa (such as the intestinal tract), dimeric IgA secreted by local plasma cells is transported
Marija Zaric et al.
Nature communications, 10(1), 2214-2214 (2019-05-19)
CD8+ T cells provide a critical defence from pathogens at mucosal epithelia including the female reproductive tract (FRT). Mucosal immunisation is considered essential to initiate this response, however this is difficult to reconcile with evidence that antigen delivered to skin
Goro Matsuzaki et al.
Immunobiology, 224(3), 440-448 (2019-02-24)
Mycobacterial antigen-specific CD4+ Th1 cells have pivotal role in protective immunity against mycobacterial infections including pulmonary tuberculosis. In the course of the infection, Th1 cells differentiate in the lung-draining lymph nodes and migrate into the infected lung. Chemokine receptors on
Kay D Hewit et al.
The Journal of biological chemistry, 289(18), 12330-12342 (2014-03-20)
The atypical chemokine receptor, ACKR2 is a pivotal regulator of chemokine-driven inflammatory responses and works by binding, internalizing, and degrading inflammatory CC-chemokines. ACKR2 displays promiscuity of ligand binding and is capable of interacting with up to 14 different inflammatory CC-chemokines.
Milena Fioravante et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(6), 1126-1137 (2019-02-11)
In experimental obesity, the hypothalamus is affected by an inflammatory response activated by dietary saturated fats. This inflammation is triggered as early as one day after exposure to a high-fat diet, and during its progression, there is recruitment of inflammatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service