Skip to Content
Merck
All Photos(9)

Key Documents

HPA013136

Sigma-Aldrich

Anti-SCG5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neuroendocrine protein 7B2 precursor, Anti-Pituitary polypeptide, Anti-Secretogranin V, Anti-Secretogranin-5, Anti-Secretory granule endocrine protein I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

EGLQHLGPFGNIPNIVAELTGDNIPKDFSEDQGYPDPPNPCPVGKTADDGCLENTPDTAEFSREFQLHQHLFDPEHDYPGLGKWNKKLLYEKMKGGERRKRRSVNPYLQGQRLDNVVAKKSV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCG5(6447)

General description

Secretogranin-5 (SCG5) is present in cells which contain secretory granules, such as neurons and endocrine cells. The gene encoding this protein is located on chromosome 15q13-q14.

Immunogen

Neuroendocrine protein 7B2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Secretogranin-5 (SCG5) functions as an anti-aggregation chaperone which is associated with many neurodegenerative diseases like Alzheimer disease. In vitro, it inhibits fibrillation and formation of amyloid-β (Aβ) and α-synuclein aggregates. SCG5 also plays a role as a molecular chaperone for proprotein convertase subtilisin/kexin type 2 (PCSK2) and prevents its premature activation in the secretory pathway. SCG5 also has an important role in pituitary hormone secretion. It may be useful as a marker in human lung cancer diagnosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71593

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ikuo Kobayashi et al.
Endocrine, 22(3), 285-292 (2004-01-08)
Recent studies have shown that 7B2 and the neuroendocrine- specific proconvertase PC2 have important roles in pituitary cell proliferation and hormone secretion. Studies from our laboratory have also shown that TGFb1 regulates anterior pituitary cell proliferation and hormone secretion. To
A J Roebroek et al.
Cancer research, 49(15), 4154-4158 (1989-08-01)
The protein designated 7B2 is a recently discovered pituitary polypeptide which is selectively expressed in cells containing secretory granules, such as neurons and endocrine cells. Northern blot analysis of 7B2 gene expression in small cell lung carcinoma (SCLC) cell lines
A J Roebroek et al.
Cytogenetics and cell genetics, 50(2-3), 158-160 (1989-01-01)
Genetic sequences encoding the novel pituitary polypeptide 7B2 were isolated from a human pituitary cDNA library. Hybridization analysis of a panel of human x mouse cell hybrids with a 7B2 cDNA probe indicated that the locus for the human 7B2
J A Braks et al.
Cell, 78(2), 263-273 (1994-07-29)
The neuroendocrine polypeptide 7B2 is a highly conserved secretory protein selectively present in prohormone-producing cells equipped with a regulated secretory pathway. We find that the amino-terminal half of 7B2 is distantly related to chaperonins, a subclass of molecular chaperones. When
Michael Helwig et al.
The Journal of biological chemistry, 288(2), 1114-1124 (2012-11-23)
Neurodegenerative diseases such as Alzheimer (AD) and Parkinson (PD) are characterized by abnormal aggregation of misfolded β-sheet-rich proteins, including amyloid-β (Aβ)-derived peptides and tau in AD and α-synuclein in PD. Correct folding and assembly of these proteins are controlled by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service