Skip to Content
Merck
All Photos(2)

Documents

HPA012716

Sigma-Aldrich

Anti-QSOX2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neuroblastoma-derived sulfhydryl oxidase, Anti-Quiescin Q6-like protein 1, Anti-Sulfhydryl oxidase 2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LWKKHNMVNGRLAGHLSEDPRFPKLQWPTPDLCPACHEEIKGLASWDEGHVLTFLKQHYGRDNLLDTYSADQGDSSEGGTLARGEEEEKRLTPPEVSHGDRDTQSVRPPGALGPRPALPESLHHSLDGKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... QSOX2(169714)

General description

The gene encoding quiescin Q6 sulfhydryl oxidase 2 (QSOX2) contains 12 exons and is localized to chromosome 9q34.3. It belongs to the sulfhydryl oxidase/Quiescin6 family. QSOX2 contains a signal sequence, a protein-disulfide-isomerase-type thioredoxin domain and yeast essential for respiration and vegetative growth protein 1 (ERV1) domain. It is expressed in the plasma and nuclear membrane.

Immunogen

Sulfhydryl oxidase 2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-QSOX2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-QSOX2 antibody produced in rabbit can also be used for immunocytochemistry.

Biochem/physiol Actions

Quiescin Q6 sulfhydryl oxidase 2 (QSOX2) catalyzes the oxidation of sulfhydryl groups of thiols in peptides and proteins to disulfides, with the reduction of molecular oxygen to hydrogen peroxide. It plays a major role in sensitization of neuroblastoma cells to interferon gamma (IFN-γ)-induced apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72164

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Wittke et al.
Cancer research, 63(22), 7742-7752 (2003-11-25)
In neuroblastoma cells, apoptotic programs can be activated by cytokines and cytostatic drugs. Apoptotic dysfunction confers resistance against therapeutic drugs and is a major complication for achieving optimal therapy response. Deregulated expression of the MYCN gene is a critical determinant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service