Skip to Content
Merck
All Photos(5)

Documents

HPA012014

Sigma-Aldrich

Anti-KRT10 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CK-10, Anti-Cytokeratin-10, Anti-K10, Anti-Keratin, type I cytoskeletal 10, Anti-Keratin-10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000-1:2500

immunogen sequence

EQLAEQNRKDAEAWFNEKSKELTTEIDNNIEQISSYKSEITELRRNVQALEIELQSQLALKQSLEASLAETEGRYCVQLSQIQAQISALEEQLQQIRAETECQNTEYQQLLDIKIRLE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT10(3858)

Immunogen

Keratin, type I cytoskeletal 10 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KRT10 (Keratin 10) is a 56.5kDa intermediate filament chain belonging to the type I keratin family. It is expressed in differentiated suprabasal cell layers of stratified epidermal cells. It is mainly involved in the formation of cytokeratin filament network to the nucleolus. Mutations in the gene cause a rare genodermatosis disorder termed as ichthyosis with confetti (IWC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70291

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iris Spoerri et al.
JAMA dermatology, 151(1), 64-69 (2014-09-12)
Ichthyosis with confetti (IWC) is a genodermatosis caused by dominant negative mutations in the gene encoding keratin 10 (KRT10). We investigated clinical and genetic details of a substantial number of patients with IWC in order to define major and minor
M Y Darmon et al.
Molecular biology reports, 12(4), 277-283 (1987-01-01)
We present here the nucleotide sequence of a 1700 bp-long cDNA encoding human epidermal keratin No. 10 (56.5 kDa). cDNA clones of the acidic keratin family were first isolated from a pBR322 human epidermal cDNA library by hybridization with a
Keith A Choate et al.
Science (New York, N.Y.), 330(6000), 94-97 (2010-08-28)
Somatic loss of wild-type alleles can produce disease traits such as neoplasia. Conversely, somatic loss of disease-causing mutations can revert phenotypes; however, these events are infrequently observed. Here we show that ichthyosis with confetti, a severe, sporadic skin disease in
X M Zhou et al.
The Journal of biological chemistry, 263(30), 15584-15589 (1988-10-25)
We present the complete amino acid sequence of the human keratin 10 (type I) intermediate filament chain expressed in terminally differentiated epidermal cells. Comparisons of this sequence with its mouse and bovine counterparts allow us to describe structural features of
Marisa Meloni et al.
International journal of cosmetic science, 43(2), 235-245 (2021-01-21)
A 3D reconstructed human epidermis (RHE) model colonized with specific microbial strains was developed to model the complex interactions between strains of the human scalp hair. Reconstructed human epidermis was colonized with Cutibacterium acnes and Malassezia restricta for 72 h. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service