Skip to Content
Merck
All Photos(8)

Key Documents

HPA011911

Sigma-Aldrich

Anti-DSC2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Desmocollin-2 precursor, Anti-Desmocollin-3, Anti-Desmosomal glycoprotein II and III

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

IGVVNEAPFSREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLDREAETIKNGIYNITVLASDQGGRTCTGTLGIILQDVNDNS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DSC2(1824)

Immunogen

Desmocollin-2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DSC2 (Desmocollin-2) is a transmembrane cadherin glycoprotein present in the desmosomal cell-cell adhesion structure. It is a member of the desmosomal cadherin family and is associated with tumor progression. It mainly performs in the cell-cell adhesion mechanism. Studies have been reported regarding the involvement of DSC2 in the regulation of cell invasiveness by controlling the cell-cell attachment and cytoskeleton rearrangement. Its expression has been found in several types of cancers such as colorectal, pancreatic, gastric, lung and urothelial cancer. Mutation in the DSC2 gene causes an inherited cardiac disease, arrhythmogenic right ventricular dysplasia/cardiomyopathy (ARVD/C) characterized by myocardial atrophy, fibro-fatty replacement, and a high risk of ventricular arrhythmias.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72495

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Brodehl et al.
Journal of molecular and cellular cardiology, 141, 17-29 (2020-03-24)
We aimed to unravel the genetic, molecular and cellular pathomechanisms of DSC2 truncation variants leading to arrhythmogenic cardiomyopathy (ACM). We report a homozygous 4-bp DSC2 deletion variant c.1913_1916delAGAA, p.Q638LfsX647hom causing a frameshift carried by an ACM patient. Whole exome sequencing
Wang-Kai Fang et al.
Molecular medicine reports, 10(5), 2358-2364 (2014-08-15)
Desmocollin‑2 (DSC2), a transmembrane glycoprotein belonging to the desmosomal cadherin family, has been found to be differentially expressed in several types of cancer and to be involved in tumor progression. The tumor metastasis suppressing property of DSC2 in esophageal squamous
Elisabetta Lazzarini et al.
Human mutation, 36(4), 403-410 (2015-02-14)
Arrhythmogenic cardiomyopathy (ACM) is an inherited cardiac disease characterized by myocardial atrophy, fibro-fatty replacement, and a high risk of ventricular arrhythmias that lead to sudden death. In 2009, genetic data from 57 publications were collected in the arrhythmogenic right ventricular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service