Skip to Content
Merck
All Photos(6)

Key Documents

HPA011389

Sigma-Aldrich

Anti-NAPG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gamma-soluble NSF attachment protein, Anti-N-ethylmaleimide-sensitive factor attachment protein gamma, Anti-SNAP-gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HLAKAEKYLKTGFLKWKPDYDSAASEYGKAAVAFKNAKQFEQAKDACLREAVAHENNRALFHAAKAYEQAGMMLKEMQKLPEAVQLIEKASMMYLENGTPDTAAMALERAGKLIENVDPEKAVQLYQQTANVFENEERLRQAVELLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAPG(8774)

Immunogen

Gamma-soluble NSF attachment protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NAPG (N-ethylmaleimide-sensitive factor attachment protein, γ) is a soluble N-ethylmaleimide-sensitive fusion (NSF)-attachment protein (SNAPs). It is associated with several cellular processes related to neurotransmission in the central nervous system. It is also involved in the SNAP receptor (SNARE) activity, vesicle docking and membrane fusion in platelets. Reports suggest that NAPG may be involved in bipolar disorder and schizophrenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71573

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NAPG may not be a common risk gene shared by bipolar disorder and schizophrenia in Chinese population.
Xing-Wang Li et al.
Psychiatric genetics, 22(2), 107-108 (2011-11-25)
P P Lemons et al.
Blood, 90(4), 1490-1500 (1997-08-15)
To further characterize the molecular mechanisms of platelet function, we have sought to identify some of the proteins that mediate the secretory events of the platelet release reaction. We report that platelets contain the general elements of the membrane transport
Xingwang Li et al.
Neuroscience letters, 457(3), 159-162 (2009-05-12)
Bipolar disorder is a mental health problem throughout the world. Chromosome 18p11 has been identified by several studies as a susceptiblilty region for bipolar disorder and NAPG, located on 18p11, has been suggested as being associated with bipolar disorder in
Andrew E Weller et al.
Psychiatric genetics, 16(1), 3-8 (2006-01-06)
A number of studies have implicated the chromosome 18p11 region as a susceptibility region for bipolar disorder. The gene encoding gamma-SNAP (NAPG), one of three soluble N-ethylmaleimide-sensitive fusion (NSF)-attachment proteins (SNAPs), is located in the 18p11 region and is thought

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service