Skip to Content
Merck
All Photos(6)

Key Documents

HPA011154

Sigma-Aldrich

Anti-GCNT3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Glucosaminyl(N-acetyl) transferase 3, mucin type antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RLKCDSDHLGLESRESQSQYCRNILYNFLKLPAKRSINCSGVTRGDQEAVLQAILNNLEVKKKREPFTDTHYLSLTRDCEHFKAERKFIQFPLSKEEVEFPIAYSMVIHEKIENFER

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCNT3(9245)

General description

GCNT3 (glucosaminyl (N-acetyl) transferase 3, mucin type) is the mucin type member of core 2 β-1,6-N-acetylglucosaminyltransferase (C2GnT) family, which has three members. This gene is located on human chromosome 15q21.3, spans ~8.26kb, and has two introns and three exons. It is predominantly expressed in mucous-secreting tissues, such as small and large intestines, trachea, and stomach as well as thyroid. It is also expressed in pancreas, kidney and colon. It is a type II transmembrane protein.

Immunogen

Glucosaminyl(N-acetyl) transferase 3, mucin type recombinant protein epitope signature tag (PrEST)

Application

Anti-GCNT3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GCNT3 (glucosaminyl (N-acetyl) transferase 3, mucin type) is responsible for the production of the three β6GlcNAc branches on secreted mucins. Not only does it catalyze the synthesis of core 2 branches, it also catalyzes the synthesis of core 4 and 1 branches. Its expression is suppressed by epidermal growth factor, and enhanced by all-trans retinoic acid and T helper 2 (Th2) cytokines such as IL-13. These cis-regulatory elements, therefore, have potential as therapeutic targets for disorders with mucus hyper-secretion. GCNT3 is responsible for the modification of O-glycans and integrin signaling, which in turn affect cell adhesion and migration and tumorigenesis. This gene is under-expressed in majority of colorectal cancer patients. It has potential to determine the relapse risk in early stage colon cancer patients, and might also help to determine the response to chemotherapy in the same.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72004

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sjoerd van der Post et al.
Molecular & cellular proteomics : MCP, 13(9), 2277-2287 (2014-06-04)
The colonic epithelium is a highly dynamic system important for the regulation of ion and water homeostasis via absorption and secretion and for the maintenance of a protective barrier between the outer milieu and the inside of the body. These
M-C Huang et al.
Oncogene, 25(23), 3267-3276 (2006-01-19)
Changes in carbohydrates on the cell surface are associated with tumor malignancy. The mucin-type core 2 beta-1,6-N-acetylglucosaminyltransferase (C2GnT-M) is highly expressed in the gastrointestinal tract and catalyses the formation of core 2, core 4, and blood group I branches on
Shuhua Tan et al.
American journal of respiratory cell and molecular biology, 36(6), 737-745 (2007-02-17)
Mucin glycan is the primary determinant of mucin functions. These functions are expanded by three branch structures, including core 2, core 4, and blood group I, which are synthesized by core 2 beta1,6 N-acetylglucosaminyltransferase-M (C2GnT-M). Alteration of C2GnT-M gene expression
T Schwientek et al.
The Journal of biological chemistry, 274(8), 4504-4512 (1999-02-13)
A novel human UDP-GlcNAc:Gal/GlcNAcbeta1-3GalNAcalpha beta1, 6GlcNAc-transferase, designated C2/4GnT, was identified by BLAST analysis of expressed sequence tags. The sequence of C2/4GnT encoded a putative type II transmembrane protein with significant sequence similarity to human C2GnT and IGnT. Expression of the
Lara P Fernández et al.
Scientific reports, 8(1), 8485-8485 (2018-06-02)
Glycosyltransferase enzyme GCNT3, has been proposed as a biomarker for prognosis in colorectal cancer (CRC). Our study goes in depth into the molecular basis of GCNT3 role in tumorigenesis and drug resistance, and it explores its potential role as biomarker

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service