Skip to Content
Merck
All Photos(7)

Key Documents

HPA011140

Sigma-Aldrich

Anti-GALNTL5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GALNT15, Anti-GalNAc-T5L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HCEVNRVWLEPLLHAIAKDPKMVVCPLIDVIDDRTLEYKPSPLVRGTFDWNLQFKWDNVFSYEMDGPEGSTKPIRSPAMSGGIFAIRRHYFNEIGQYDKDMD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

GALNTL5 (polypeptide N-acetylgalactosaminyltransferase-like 5) is a part of the polypeptide N-acetylgalactosamine-transferase (pp-GalNAc-T) family of proteins. It does not contain the conserved lectin domain at the C-terminal, unlike other members of this family, but it has the conserved catalytic domains of pp-GalNAc-T family. One isoform of this protein is exclusively expressed in human testis. This gene is localized to human chromosome 7q36.1.

Immunogen

polypeptide N-acetylgalactosaminyltransferase-like 5

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GALNTL5 (polypeptide N-acetylgalactosaminyltransferase-like 5) is responsible for the O-linked glycosylation of proteins. Heterozydous deletion in this gene is associated with asthenozoospermia. Therefore, GALNTL5 might be involved in spermatogenesis and maturation of sperms.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71763

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of ethnic differences in the prevalence of chromosomal number aberrations of luminal and triple-negative breast cancers associated with risk of recurrence.
Brewster AM
Journal of Clinical Oncology, 28(15), 1526-1526 (2010)
Bert van der Zwaag et al.
PloS one, 4(5), e5324-e5324 (2009-06-06)
The recent identification of copy-number variation in the human genome has opened up new avenues for the discovery of positional candidate genes underlying complex genetic disorders, especially in the field of psychiatric disease. One major challenge that remains is pinpointing
Nobuyoshi Takasaki et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(3), 1120-1125 (2014-01-09)
For normal fertilization in mammals, it is important that functionally mature sperm are motile and have a fully formed acrosome. The glycosyltransferase-like gene, human polypeptide N-acetylgalactosaminyltransferase-like protein 5 (GALNTL5), belongs to the polypeptide N-acetylgalactosamine-transferase (pp-GalNAc-T) gene family because of its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service