Skip to Content
Merck
All Photos(4)

Key Documents

HPA010607

Sigma-Aldrich

Anti-AFP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-1-Fetoprotein antibody produced in rabbit, Anti-α-Fetoglobulin antibody produced in rabbit, Anti-α-Fetoprotein precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
recombinant expression
RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FAQFVQEATYKEVSKMVKDALTAIEKPTGDEQSSGCLENQLPAFLEELCHEKEILEKYGHSDCCSQSEEGRHNCFLAHKKPTPASIPLFQVPEPVTSCEAYEEDRETFMNKFIYEIARRHPFLYAPTILLWAAR

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFP(174)

General description

AFP (α fetoprotein) is a 70kDa serum glycoprotein, and is produced during gestation from fetal liver and yolk sac.

Immunogen

α-Fetoprotein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-AFP antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

AFP (α fetoprotein) levels decrease by the second year of birth and its ectopic expression is linked with tumors, such as hepatobastoma, hepatocellular carcinoma (HCC) and yolk sac tumors. Maximum AFP-expressing tumors are of stomach, bile duct or pancreatic origin. AFP-expressing rectal cancer is extremely rare and is usually metastasized to liver with poor prognosis. In patients with chronic hepatitis C, high levels of this protein are linked with increased risk of developing HCC. In patients with metastatic gastric cancer (GC), follow-up of AFP concentrations can help determine early treatment response. For early and intermediate stages of HCC, the addition of this protein and ascites in the BCLC Barcelona Clinic Liver Cancer (BCLC) staging can improvise prognosis determination.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86803

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Shimamoto et al.
PloS one, 9(11), e112900-e112900 (2014-11-13)
Werner syndrome (WS) is a premature aging disorder characterized by chromosomal instability and cancer predisposition. Mutations in WRN are responsible for the disease and cause telomere dysfunction, resulting in accelerated aging. Recent studies have revealed that cells from WS patients
Asmaa I Gomaa et al.
World journal of gastroenterology, 21(18), 5654-5662 (2015-05-20)
To assess how ascites and alpha-fetoprotein (AFP) added to the Barcelona Clinic Liver Cancer (BCLC) staging predict hepatocellular carcinoma survival. The presence of underlying cirrhosis, ascites and encephalopathy, Child-Turcotte-Pugh (CTP) score, the number of nodules, and the maximum diameter of
Hiroyuki Anzai et al.
World journal of surgical oncology, 13, 180-180 (2015-05-13)
Alpha-fetoprotein (AFP)-producing rectal cancer is very rare, and this type of cancer frequently metastasizes to the liver with a poor prognosis. To date, only 11 cases of AFP-producing colorectal cancer have been reported. A 41-year-old woman was first presented to
Koji Takayama et al.
World journal of gastroenterology, 21(15), 4696-4706 (2015-04-29)
To investigate the impact of telaprevir-based triple therapy on the serum alpha-fetoprotein (AFP) level of chronic hepatitis C patients. A total of 210 patients with chronic hepatitis C genotype 1 of high viral load (baseline serum hepatitis C virus RNA
Ali Murat Tatli et al.
Asian Pacific journal of cancer prevention : APJCP, 16(5), 2003-2007 (2015-03-17)
Elevated serum alpha-fetoprotein (AFP) levels in adults are considered abnormal. This parameter is used mostly in the diagnosis and follow-up of hepatocellular carcinomas and yolk sac tumors. Among the other rare tumors accompanied with elevated serum AFP levels, gastric cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service