Skip to Content
Merck
All Photos(8)

Key Documents

HPA010568

Sigma-Aldrich

Anti-KIF11 antibody produced in rabbit

enhanced validation

affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution

Synonym(s):

Anti-Kinesin-like protein 1, Anti-Kinesin-like protein KIF11, Anti-Kinesin-like spindle protein HKSP, Anti-Kinesin-related motor protein Eg5, Anti-TRIP-5, Anti-Thyroid receptor-interacting protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ALIKEYTEEIERLKRDLAAAREKNGVYISEENFRVMSGKLTVQEEQIVELIEKIGAVEEELNRVTELFMDNKNELDQCKSDLQNKTQELETTQKHLQETKLQLVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIF11(3832)

General description

KIF11 (kinesin family member 11) gene is localized to human chromosome 10q23.33, which codes for a protein composed of 1056 amino acids. Its N-terminal contains the motor domain, a microtubule-binding domain, an ATP-binding domain, along with three coiled coil domains. It contains BimC domain at its C-terminal.

Immunogen

Kinesin-like protein KIF11 recombinant protein epitope signature tag (PrEST)

Application

Anti-KIF11 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

KIF11 (kinesin family member 11) forms homotetrameric kinesin complexes. Mutations in this gene are linked with microcephaly, lymphedema and chorioretinal dysplasia syndrome (MLCRD), and chorioretinal dysplasia, microcephaly and mental retardation syndrome (CDMMR). This protein crosslinks two anti-parallel microtubules and moves to plus-ends of both the microtubules, thus, promoting the separation of spindle poles in a mitotic cell. It might also be involved in transport in cytoplasm. In prostate cancer patients, the nuclear expression of this protein acts as a marker to predict the aggressiveness of the tumor and the response to docetaxel.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71070

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Kowalewski et al.
Polish journal of pathology : official journal of the Polish Society of Pathologists (2022-08-19)
Unresectable renal cell carcinoma continues to be a great challenge due to our limited understanding of its underlying pathophysiology. We explored the relationship between KIF11 protein expression and the clinical courses of clear cell renal cell carcinoma (ccRCC) using a tissue microarray.
Kayo Daigo et al.
International journal of oncology, 52(1), 155-165 (2017-11-09)
Oral cancer has a high mortality rate, and its incidence is increasing gradually worldwide. As the effectiveness of standard treatments is still limited, the development of new therapeutic strategies is eagerly awaited. Kinesin family member 11 (KIF11) is a motor protein required
Matthieu J Schlögel et al.
Orphanet journal of rare diseases, 10, 52-52 (2015-05-03)
Microcephaly with or without chorioretinopathy, lymphedema, or mental retardation syndrome (MCLMR) is a rare autosomal dominant disorder with variable expressivity. It is characterized by mild-to-severe microcephaly, often associated with intellectual disability, ocular defects and lymphedema. It can be sporadic or
Distinct dynein complexes defined by DYNLRB1 and DYNLRB2 regulate mitotic and male meiotic spindle bipolarity.
He, et al.
Nature Communications, 14, 1715-1715 (2023)
T Kuga et al.
Oncogenesis, 3, e94-e94 (2014-03-19)
The majority of human cancer shows chromosomal instability (CIN). Although the precise mechanism remains largely uncertain, proper progression of mitosis is crucial. B-type lamins were suggested to be components of the spindle matrix of mitotic cells and to be involved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service