Skip to Content
Merck
All Photos(2)

Key Documents

HPA008703

Sigma-Aldrich

Anti-EVC antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DWF-1 antibody produced in rabbit, Anti-Ellis-van Creveld syndrome protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

GVMDLLEAQLETQLQEAEQNFISELAALARVPLAESKLLPAKRGLLEKPLRTKRKKPLPQERGDLGVPNNEDLASGDQTSGSLSSKRLSQQESEAGDSGNSKKMLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EVC(2121)

Looking for similar products? Visit Product Comparison Guide

General description

EVC (Ellis van Creveld) gene is localized to the human chromosome 4p16, and is composed of 21 exons, spanning 120kb. The encoded protein is composed of 992 amino acids. This gene, along with EVC2 gene, forms the EVC gene family.

Immunogen

Ellis-van Creveld syndrome protein recombinant protein epitope signature tag (PrEST)

Application

Anti-EVC antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Mutations in EVC (Ellis van Creveld) gene are responsible for EVC syndrome, which is an autosomal recessive disorder and is characterized by postaxial polydactyly, short ribs and limbs and dysplastic teeth and nails. This gene is over-expressed in adult T-cell leukemia (ATL) and human T-cell leukemia virus type I (HTLV-1)-infected cells. Thus, it has potential as a marker and therapeutic target in HTLV-1-infected T-cells. This gene plays a key role in hedgehog signaling, especially during the growth of cartilage plate. It is localized at the basal body and cilia membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71642

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hakan Ulucan et al.
BMC medical genetics, 9, 92-92 (2008-10-25)
Ellis-van Creveld (EvC) syndrome is characterized by short limbs, short ribs, postaxial polydactyly, dysplastic nails and teeth and is inherited in an autosomal recessive pattern. We report a family with complex septal cardiac defects, rhizomelic limb shortening, and polydactyly, without
Helen J Blair et al.
BMC biology, 9, 14-14 (2011-03-02)
Evc is essential for Indian Hedgehog (Hh) signalling in the cartilage growth plate. The gene encoding Evc2 is in close proximity in divergent orientation to Evc and mutations in both human genes lead to the chondrodysplasia Ellis-van Creveld syndrome. Bioinformatic
Honghao Zhang et al.
Genesis (New York, N.Y. : 2000), 53(9), 612-626 (2015-07-30)
Ellis-van Creveld (EvC) syndrome (OMIM 225500) is an autosomal recessive disease characterized with chondrodysplastic dwarfism in association with abnormalities in oral cavity. Ciliary proteins EVC and EVC2 have been identified as causative genes and they play an important role on
Honghao Zhang et al.
PLoS genetics, 12(12), e1006510-e1006510 (2016-12-28)
Ellis-van Creveld (EvC) syndrome is a skeletal dysplasia, characterized by short limbs, postaxial polydactyly, and dental abnormalities. EvC syndrome is also categorized as a ciliopathy because of ciliary localization of proteins encoded by the two causative genes, EVC and EVC2
Ryutaro Takahashi et al.
Cancer science, 105(9), 1160-1169 (2014-07-06)
One of the hallmarks of cancer, global gene expression alteration, is closely associated with the development and malignant characteristics associated with adult T-cell leukemia (ATL) as well as other cancers. Here, we show that aberrant overexpression of the Ellis Van

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service