Skip to Content
Merck
All Photos(2)

Key Documents

HPA008192

Sigma-Aldrich

Anti-TBX20 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-T-box protein 20, Anti-T-box transcription factor TBX20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

DSPFTGEQLLKQMVSFEKVKLTNNELDQHGHIILNSMHKYQPRVHIIKKKDHTASLLNLKSEEFRTFIFPETVFTAVTAYQNQLITKLKIDSNPFAKGFRDSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBX20(57057)

General description

T-box transcription factor (TBX20) belongs to the T-box transcription factor family. It is expressed in the developing hearts of embryos in vertebrates and invertebrates. TBX20 contains a DNA-binding domain called the T-box domain and the gene encoding this protein is present on chromosome 7p14-p15.

Immunogen

T-box transcription factor TBX20 recombinant protein epitope signature tag (PrEST)

Application

Anti-TBX20 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence[1] and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

T-box transcription factor (TBX20) activates cardiac gene expression by interacting with transcription factors NK2 homeobox 5 (Nkx2-5), GATA binding protein 4 (GATA4) and GATA binding protein 5 (GATA5). During endocardial differentiation, it specifically interacts with GATA5. TBX20 and other T-box genes and factors interact repressively. This is responsible for splitting of the myocardium into the chamber and non-chamber. It is an important event upon which all the subsequent morphogenesis activities depend. Based on the particular isoform expressed and cellular requirements, TBX20 can regulate transcription of target genes both positively and negatively. Its T-box DNA-binding domain associates specifically with the consensus DNA half site sequence defined for the brachyury (T) protein. TBX20 also interacts with homeodomain factor Nkx2-5 and zinc finger factor GATA4.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70963.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the human TBX20 gene, a new member of the T-Box gene family closely related to the Drosophila H15 gene.
M Meins
Genomics, 67(3), 317-332 (2000)
Fiona A Stennard et al.
Development (Cambridge, England), 132(10), 2451-2462 (2005-04-22)
The genetic hierarchies guiding lineage specification and morphogenesis of the mammalian embryonic heart are poorly understood. We now show by gene targeting that murine T-box transcription factor Tbx20 plays a central role in these pathways, and has important activities in
Fiona A Stennard et al.
Developmental biology, 262(2), 206-224 (2003-10-11)
Tbx20 is a member of the T-box transcription factor family expressed in the forming hearts of vertebrate and invertebrate embryos. We report here analysis of Tbx20 expression during murine cardiac development and assessment of DNA-binding and transcriptional properties of Tbx20
Yuelong Liu et al.
Human molecular genetics, 23(8), 2145-2156 (2013-12-03)
Haploinsufficiency for CHD7, an ATP-dependent nucleosome remodeling factor, is the leading cause of CHARGE syndrome. While congenital heart defects (CHDs) are major clinical features of CHARGE syndrome, affecting >75% of patients, it remains unclear whether CHD7 can directly regulate cardiogenic
Mengcheng Shen et al.
STAR protocols, 4(2), 102256-102256 (2023-04-29)
Cardiac pericytes are a critical yet enigmatic cell type within the coronary microvasculature. Since primary human cardiac pericytes are not readily accessible, we present a protocol to generate them from human induced pluripotent stem cells (hiPSCs). Our protocol involves several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service