Skip to Content
Merck
All Photos(4)

Key Documents

HPA008026

Sigma-Aldrich

Anti-STK40 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SINK-homologous serine/threonine-protein kinase, Anti-Serine/threonine-protein kinase 40, Anti-SgK495, Anti-Sugen kinase 495

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AKALGSGISGNNAKRAGPFILGPRLGNSPVPSIVQCLARKDGTDDFYQLKILTLEERGDQGIESQEERQGKMLLHTEYSLLSLLHTQDGVVHHHGLFQDRTCEIVEDTESSRMVKKMKKRICLVLDCLCAHDFSDKTADL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STK40(83931)

Immunogen

Serine/threonine-protein kinase 40 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

STK40 (serine/threonine kinase 40) is a putative serine/threonine kinase that negatively regulates TNF-induced NF-κB activation and p53-induced transcription. It induces extra embryonic endoderm differentiation by linking the pluripotency factor Oct4 to the Erk/MAPK pathway. It is also an essential factor in the maturation and function of alveolar epithelium, and its deletion causes respiratory failure and death of mice during birth .

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71530

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyao Yu et al.
The Journal of biological chemistry, 288(8), 5342-5352 (2013-01-08)
STK40 is a putative serine/threonine kinase and was shown to induce extraembryonic endoderm differentiation from mouse embryonic stem cells. However, little is known about its physiological function in vivo. Here, we generate Stk40 knock-out mice and demonstrate that loss of
Jun Huang et al.
Biochemical and biophysical research communications, 309(4), 774-778 (2003-09-19)
SINK is a p65-interacting protein that inhibits PKAc-induced phosphorylation of p65 and NF-kappaB transcriptional competence. We identified a SINK-homologous serine/threonine kinase SHIK. SHIK is ubiquitously expressed and is localized in the cytoplasm. Overexpression of SHIK inhibits TNF-triggered NF-kappaB activation in
Lingjie Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(4), 1402-1407 (2010-01-19)
Self-renewal and differentiation of embryonic stem cells (ESCs) are controlled by intracellular transcriptional factors and extracellular factor-activated signaling pathways. Transcription factor Oct4 is a key player maintaining ESCs in an undifferentiated state, whereas the Erk/MAPK pathway is known to be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service