Skip to Content
Merck
All Photos(1)

Documents

HPA007978

Sigma-Aldrich

Anti-PTH1R antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PTH/PTHr receptor, Anti-PTH/PTHrP type I receptor, Anti-PTH1 Receptor, Anti-Parathyroid hormone 1 receptor, Anti-Parathyroid hormone/parathyroid hormone-related peptide receptor precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RWTLALDFKRKARSGSSSYSYGPMVSHTSVTNVGPRVGLGLPLSPRLLPTATTNGHPQLPGHAKPGTPALETLETTPPAMAAPKDDGFLNGSCSGLDEEASGPERPPALLQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTH1R(5745)

General description

PTH1R (parathyroid hormone 1 receptor) gene encodes a member of the G-protein coupled receptor family B or secretin family GPCRs. These receptors have an N-terminal extracellular domain (ECD) spanning a length of 160 amino acids and a 7-TM domain.

Immunogen

Parathyroid hormone/parathyroid hormone-related peptide receptor Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PTH1R (parathyroid hormone 1 receptor) gene encodes a protein that serves as a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH). PTH is involved in the maintenance of calcium and phosphate homeostasis by activating PTH1R expressed in bone and kidney tissues. PTHLH is a paracrine factor that regulates bone development and is also found to be secreted by tumors that cause humoral hypercalcemia of malignancy. The activity of PTH1R is mediated by G proteins that are involved in the activation of adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system. Defects in this gene are associated with Jansen′s metaphyseal chondrodysplasia (JMC), chondrodysplasia Blomstrand type (BOCD), and enchodromatosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71228

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murat Bastepe et al.
The Journal of clinical endocrinology and metabolism, 89(7), 3595-3600 (2004-07-09)
A novel heterozygous PTH/PTHrP receptor missense mutation (T410R) was identified in a male and his two sons who are all affected by a less severe form of Jansen's metaphyseal chondrodysplasia (JMC). JMC is a rare disorder that is typically characterized
Jiawei Zhang et al.
Journal of cellular physiology, 234(7), 11900-11911 (2018-12-26)
Dental follicle cells (DFCs) activate and recruit osteoclasts for tooth development and tooth eruption, whereas DFCs themselves differentiate into osteoblasts to form alveolar bone surrounding tooth roots through the interaction with Hertwig's epithelial root sheath (HERS). Also during tooth development
E Schipani et al.
The Journal of clinical endocrinology and metabolism, 84(9), 3052-3057 (1999-09-16)
Two heterozygous PTH/PTH-related peptide (PTHrP) receptor missense mutations were previously identified in patients with Jansen's metaphyseal chondrodysplasia (JMC), a rare form of short limb dwarfism associated with hypercalcemia and normal or undetectable levels of PTH and PTHrP. Both mutations, H223R
Phillip W Hwang et al.
Scientific reports, 9(1), 8299-8299 (2019-06-06)
The spontaneously immortalized murine calvarial cell line MC3T3-E1 and its derivative subclones are widely used models of osteoblast biology. Many investigators have reported conflicting data under seemingly similar experimental conditions, though the specific subclone studied is often not specified. The
E Schipani et al.
The New England journal of medicine, 335(10), 708-714 (1996-09-05)
An activating mutation of the receptor for parathyroid hormone (PTH) and parathyroid hormone-related peptide (PTHrP) was recently found in a patient with Jansens's metaphyseal chondrodysplasia, a rare form of short-limbed dwarfism associated with hypercalcemia and normal or low serum concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service