Skip to Content
Merck
All Photos(3)

Key Documents

HPA007925

Sigma-Aldrich

Anti-YWHAB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-YWHAA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ELSNEERNLLSVAYKNVVGGRRSAWRVISSIEQKTDTSDKKLQLIKDYREKVESELRSICTTVLELLDKYLIANATNPESKVFYLKMKGDYFRY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YWHAB(7529)

Immunogen

tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

YWHAQ (tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, θ) gene encodes a protein belonging to the 14-3-3 family of proteins that are involved in signal transduction by binding to phosphorylated Ser residues via the two consensus sequences of RXXXpSXP and R(S/X)XpSXP. The functional unit is a dimer and may function in several processes including signal transduction, apoptosis, cell cycle progression and DNA replication. The encoded protein is also called as 14-3-3 θ. It may serve as a potential marker of lung cancer. This protein has been found to be upregulated in amyotrophic lateral sclerosis spinal cord. The promoter of YWHAQ gene has been found to be hypermethylated in preeclamptic placentas.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71118

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Liu et al.
Placenta, 35(12), 981-988 (2014-10-13)
Disruption of the 14-3-3 tau (YWHAQ) gene has been shown to be involved in preeclampsia (PE). The YWHAQ promoter could be differentially regulated by methylation in severe PE patients. Placental genomic DNA from patients with severe PE (n = 21)
Wenqing Qi et al.
International journal of cancer, 113(3), 359-363 (2004-09-30)
14-3-3 Proteins play important roles in a wide range of vital regulatory processes, including signal transduction, apoptosis, cell cycle progression and DNA replication. In mammalian cells, 7 14-3-3 isoforms (beta, gamma, epsilon, eta, sigma, theta and zeta) have been identified
A Malaspina et al.
Journal of neurochemistry, 75(6), 2511-2520 (2000-11-18)
We have recently isolated a 2.2-kb cDNA clone (1C5) from a human spinal cord cDNA library with partial identity to the 14-3-3 protein mRNA encoding the theta protein (YWHAQ). 14-3-3 protein transcripts are highly expressed in large projection neurones of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service