Skip to Content
Merck
All Photos(4)

Key Documents

HPA007856

Sigma-Aldrich

Anti-CCAR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CARP-1 antibody produced in rabbit, Anti-Cell cycle and apoptosis regulatory protein 1 antibody produced in rabbit, Anti-Cell division cycle and apoptosis regulator protein 1 antibody produced in rabbit, Anti-Death inducer with SAP domain antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KDVEENVGLIVYNGAMVDVGSLLQKLEKSEKVRAEVEQKLQLLEEKTDEDEKTILNLENSNKSLSGELREVKKDLSQLQENLKISENMNLQFENQMNKTIRNLSTVMDEIHTVLKKDNVKNED

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCAR1(55749)

Immunogen

Cell division cycle and apoptosis regulator protein 1 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

CCAR1 (cell division cycle and apoptosis regulator 1) gene encodes a co-activator protein that associates with and recruits components of the mediator and p160 coactivator complexes by nuclear receptors (NR) to target genes. In response to appropriate hormone, it is recruited to endogenous NR target genes. It is involved in the regulation of genes that are involved in cell proliferation. It also functions as a p53 coactivator and regulates transcription. Cellular stress induces microtubule-dependent formation of RNA stress granules via CCAR1 and AKAP350A, a multifunctional scaffolding protein. It is a perinuclear protein that mediates apoptosis signaling and activates caspase 9 and 3, thereby exhibiting pro-apoptotic action.[1] It also functions as a coactivator of androgen receptor (AR) and facilitates the formation of AR transcription complex. It functions in androgen signaling and prostate tumorigenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71479

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang Yun Ha et al.
Cancer research and treatment : official journal of Korean Cancer Association, 48(3), 1065-1073 (2015-10-30)
Cell division cycle and apoptosis regulator 1 (CCAR1) plays a dynamic role in regulation of cell growth and apoptosis by serving as a cofactor of steroid/thyroid nuclear receptors, β-catenin, and p53 in a variety of cell types including different cancer
Jeong Hoon Kim et al.
Molecular cell, 31(4), 510-519 (2008-08-30)
DNA-bound transcription factors recruit many coactivator proteins to remodel chromatin and activate transcription. The Mediator complex is believed to recruit RNA polymerase II to most protein-encoding genes. It is generally assumed that interaction of Mediator subunits with DNA-binding transcription factors
Stephanie François et al.
PloS one, 7(8), e43335-e43335 (2012-08-21)
Regeneration of muscle fibers, lost during pathological muscle degeneration or after injuries, is sustained by the production of new myofibers by means of the satellite cells. Survival of the satellite cells is a critical requirement for efficient muscle reconstitution. Necdin
Elena Kolobova et al.
Experimental cell research, 315(3), 542-555 (2008-12-17)
Recent investigations have highlighted the importance of subcellular localization of mRNAs to cell function. While AKAP350A, a multifunctional scaffolding protein, localizes to the Golgi apparatus and centrosomes, we have now identified a cytosolic pool of AKAP350A. Analysis of AKAP350A scaffolded
Woo-Young Seo et al.
Nucleic acids research, 41(18), 8526-8536 (2013-07-28)
Androgen receptor (AR), a ligand-dependent transcription factor, plays a critical role in prostate cancer onset and progression, and its transcriptional function is mediated largely by distinct nuclear receptor co-regulators. Here, we show that cell cycle and apoptosis regulator 1 (CCAR1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service