Skip to Content
Merck
All Photos(10)

Key Documents

HPA006753

Sigma-Aldrich

Anti-PITRM1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Metalloprotease 1 antibody produced in rabbit, Anti-Pitrilysin metalloproteinase 1 antibody produced in rabbit, Anti-Presequence protease, mitochondrial precursor antibody produced in rabbit, Anti-hMP1 antibody produced in rabbit, Anti-hPreP antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ELRSQQSKPQDASCLPALKVSDIEPTIPVTELDVVLTAGDIPVQYCAQPTNGMVYFRAFSSLNTLPEELRPYVPLFCSVLTKLGCGLLDYREQAQQIELKTGGMSASPHVLPDDSHMDTYEQGVLFSSLCLDRNLPDMMQLWSEI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PITRM1(10531)

Looking for similar products? Visit Product Comparison Guide

General description

PITRM1 (Pitrilysin metallopeptidase 1) is a metalloendoprotease belonging to the pitrilysin family. It comprises of a conserved catalytic domain at the N-terminal region. It is highly expressed in the muscle, heart and lesser extent in the brain, pancreas, liver, lung, and placenta.

Immunogen

Presequence protease, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PITRM1 (Pitrilysin metallopeptidase 1) acts as a mitochondrial amyloid-β peptide (Aβ)-degrading enzyme localized in the mitochondrial matrix. Its activity is controlled by Zn+2-dependent metalloprotease inhibitor. The temporal lobe of the human brain has a high tendency of Aβ accumulation and reactive oxygen species (ROS) production. In Alzheimer′s disease, the low proteolytic activity of PITRM1 in the brain mitochondria has been reported. Studies have been suggested that high ROS production results in decreased PITRM1 activity followed by Aβ accumulation. As a result, it leads to mitochondrial toxicity and neuronal death. Thus, it proves the impact of PITRM1 in the Alzheimer′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70795

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Mzhavia et al.
DNA and cell biology, 18(5), 369-380 (1999-06-09)
A novel cDNA, designated human metalloendoprotease 1 (hMP1), was identified on the basis of homology to known metalloendoproteases of the pitrilysin family. The full-length MP1 codes for a protein with an open reading frame of 1038 amino acids. The N-terminal
Nyosha Alikhani et al.
Journal of Alzheimer's disease : JAD, 27(1), 75-87 (2011-07-14)
Accumulation of amyloid-β peptide (Aβ), the neurotoxic peptide implicated in the pathogenesis of Alzheimer's disease (AD), has been shown in brain mitochondria of AD patients and of AD transgenic mouse models. The presence of Aβ in mitochondria leads to free
Catarina Moreira Pinho et al.
Neuroscience letters, 469(2), 204-208 (2009-12-08)
Several studies suggest mitochondrial dysfunction as a possible mechanism underlying the development of Alzheimer disease (AD). There is data showing that amyloid-beta (A beta) peptide is present in AD brain mitochondria. The human presequence protease (hPreP) was recently shown to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service