Skip to Content
Merck
All Photos(3)

Key Documents

HPA006738

Sigma-Aldrich

Anti-CDK9 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-2K antibody produced in rabbit, Anti-Cell division cycle 2-like protein kinase 4 antibody produced in rabbit, Anti-Cell division protein kinase 9 antibody produced in rabbit, Anti-Cyclin-dependent kinase 9 antibody produced in rabbit, Anti-Serine/threonine-protein kinase PITALRE antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HQLALISQLCGSITPEVWPNVDNYELYEKLELVKGQKRKVKDRLKAYVRDPYALDLIDKLLVLDPAQRIDSDDALNHDFFWSDPMPSDLKGMLSTHLTSMFEYLAPPRRKGSQITQQSTNQSRNPATTNQTEFE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK9(1025)

Immunogen

Cell division protein kinase 9 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CDK9 (Cyclin-dependent kinase 9) is a ~43kDa protein localized to the nucleus. It is a member of cell division cycle 2 (CDC2) kinase family. CDK9 functions as a catalytic subunit of RNA polymerase II (RNAP II) during its transcription. It forms a multiprotein complex by binding to the P-TEFb (positive transcription elongation factor b) and a cyclin subunit (cyclin T1 or cyclin T2). It plays a vital role in the transcriptional regulation. It phosphorylates the carboxyl-terminal domain (CTD) of RNAPII to accelerate the transcription elongation. It also interacts with HIV-1 Tat protein proving an involvement of this protein in AIDS.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86958

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario Failli et al.
Hepatology (Baltimore, Md.), 80(1), 55-68 (2023-09-20)
Hepatoblastoma (HB) is the predominant form of pediatric liver cancer, though it remains exceptionally rare. While treatment outcomes for children with HB have improved, patients with advanced tumors face limited therapeutic choices. Additionally, survivors often suffer from long-term adverse effects
T J Fu et al.
The Journal of biological chemistry, 274(49), 34527-34530 (1999-11-27)
Important progress in the understanding of elongation control by RNA polymerase II (RNAPII) has come from the recent identification of the positive transcription elongation factor b (P-TEFb) and the demonstration that this factor is a protein kinase that phosphorylates the
X Graña et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(9), 3834-3838 (1994-04-26)
Members of the cell division cycle 2 (CDC2) family of kinases play a pivotal role in the regulation of the eukaryotic cell cycle. In this communication, we report the isolation of a cDNA that encodes a CDC2-related human protein kinase
P Wei et al.
Cell, 92(4), 451-462 (1998-03-10)
The HIV-1 Tat protein regulates transcription elongation through binding to the viral TAR RNA stem-loop structure. We have isolated a novel 87 kDa cyclin C-related protein (cyclin T) that interacts specifically with the transactivation domain of Tat. Cyclin T is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service