Skip to Content
Merck
All Photos(9)

Key Documents

HPA006417

Sigma-Aldrich

Anti-HTATIP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-30 kDa HIV-1 TAT-interacting protein antibody produced in rabbit, Anti-HIV-1 TAT-interactive protein 2 antibody produced in rabbit, Anti-Oxidoreductase HTATIP2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

ALSKLREDFRMQNKSVFILGASGETGRVLLKEILEQGLFSKVTLIGRRKLTFDEEAYKNVNQEVVDFEKLDDYASAFQGHDVGFC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTATIP2(10553)

General description

HTATIP2 (HIV-1 Tat interactive protein 2) has a molecular weight of 30kDa, and has altered expression in lung, breast, liver, gastric, gallbladder, and colorectal cancer. It is an evolutionary conserved gene, and has a ubiquitous pattern of expression. It is a cytoplasmic protein, and the corresponding gene is localized to human chromosome 11p15.1.

Immunogen

Oxidoreductase HTATIP2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HTATIP2 (HIV-1 Tat interactive protein 2) inhibits angiogenesis and thus suppresses tumorigenesis and metastasis. This gene, along with microvessel density (MVD), can help determine the survival of hepatocellular carcinoma, with or without sorafenib treatment. In pancreatic cancer, the decreased expression of this gene leads to elevated levels of Snail family members, which eventually results in increased metastasis and invasion. Thus, down-regulation of this gene is linked with poor prognosis in patients with pancreatic ductal adenocarcinoma (PDAC). It is also linked with ovarian cancer, and might have potential in the prevention, diagnosis and treatment of the same. It is responsible for the modification in membrane phosphatidic acid (PA), thus, facilitating membrane fusion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70890

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiwei Guo et al.
International journal of cancer, 134(6), 1369-1378 (2013-09-17)
Pancreatic ductal adenocarcinoma (PDAC) is known for its aggressive growth, and is characterized by early tissue invasion and metastasis with poor prognosis. Identifying prognostic markers and delineating the underlying mechanisms that promote progression of PDAC are important for the treatment
Wen-Quan Wang et al.
Oncotarget, 5(11), 3895-3906 (2014-07-11)
Our previous studies have demonstrated that sorafenib can promote the dissemination of hepatocellular carcinoma (HCC) through downregulation of HTATIP2, a suppressor of tumor growth and metastasis that is associated with inhibition of angiogenesis. Here, we investigated the predictive values of
Yakup Kumtepe et al.
European journal of medical research, 18, 18-18 (2013-06-27)
Human HIV-1 TAT interactive protein 2 (HTATIP2/TIP30) is an evolutionarily conserved gene that is expressed ubiquitously in human tissues and some tumor tissues. This protein has been found to be associated with some gynecological cancers; as such, this study aimed
Chengliang Zhang et al.
PloS one, 6(6), e21233-e21233 (2011-07-07)
Efficient membrane fusion has been successfully mimicked in vitro using artificial membranes and a number of cellular proteins that are currently known to participate in membrane fusion. However, these proteins are not sufficient to promote efficient fusion between biological membranes
Xin Tong et al.
The American journal of pathology, 174(5), 1931-1939 (2009-04-08)
The HIV Tat-interacting protein (TIP30), also called CC3 or HTIP2, is encoded by Tip30, a putative tumor-suppressor gene located on human chromosome 11p15.1. In this study, we investigated the role of TIP30 in the progression and metastasis of lung cancer.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service