Skip to Content
Merck
All Photos(7)

Key Documents

HPA006312

Sigma-Aldrich

Anti-ISY1 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RAB43 antibody produced in rabbit, Anti-Ras-related protein Rab-41 antibody produced in rabbit, Anti-Ras-related protein Rab-43 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GKVKERRPFLASECTELPKAEKWRRQIIGEISKKVAQIQNAGLGEFRIRDLNDEINKLLREKGHWEVRIKELGGPDYGKVGPKMLDHEGKEVPGNRGYKYFGAAKDLPGVRELFEKEPLPPPRK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ISY1(57461)

Looking for similar products? Visit Product Comparison Guide

General description

ISY1 (interactor of Syf1p) is a splicing factor, which makes a part of the cell cycle and splicing complex (CSC), which acts as a spliceosome.

Immunogen

Ras-related protein Rab-43 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ISY1 (interactor of Syf1p) binds to and facilitates the premature release of the ATPase Prp16p. Thus, it is essential for proper pre-mRNA splicing. Mutations in this gene lead to cell death, especially in the absence of the protein Ntc20p. Also, double mutation of ISY1 and SYF gene result in cell cycle arrest in a temperature-dependent manner. Studies suggest that ISY1 gene is important for the optimization of the splicing process.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70755

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orna Dahan et al.
Nucleic acids research, 30(20), 4361-4370 (2002-10-18)
Previous work has identified a group of genes whose products play important roles in two seemingly unrelated processes: cell cycle progression and splicing. The products of these genes show a network of physical and genetic interactions suggestive of the existence
I Dix et al.
RNA (New York, N.Y.), 5(3), 360-368 (1999-03-27)
We have identified a novel splicing factor, Isy1p, through two-hybrid screens for interacting proteins involved in nuclear pre-mRNA splicing. Isy1p was tagged and demonstrated to be part of the splicing machinery, associated with spliceosomes throughout the splicing reactions. At least
Tommaso Villa et al.
Genes & development, 19(16), 1894-1904 (2005-08-17)
Prp16p is a DEAH-box ATPase that transiently associates with the spliceosome to promote the structural transition required for the second chemical step. Yeast strains carrying the cold-sensitive allele prp16-302 stall the release of Prp16p at low temperatures, yet splice precursors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service