Skip to Content
Merck
All Photos(4)

Documents

HPA005853

Sigma-Aldrich

Anti-TRAK1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-106 kDa O-GlcNAc transferase-interacting protein, Anti-Trafficking kinesin-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ELEDKYAECMEMLHEAQEELKNLRNKTMPNTTSRRYHSLGLFPMDSLAAEIEGTMRKELQLEEAESPDITHQKRVFETVRNINQVVKQRSLTPSPMNIPGSNQSSAMNSLLSSCVSTPRSSFYGSDIGNVVLDNKTNSIILET

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRAK1(22906)

Immunogen

Trafficking kinesin-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Trafficking kinesin-binding protein 1 is a protein encoded by the TRAK1 gene in humans. It is referred as MILT1 and OIP106. This gene may target O-GlcNAc transferase (OGT) to transcriptional complexes for glycosylation of transcriptional proteins (such as RNA polymerase II and transcription factors). It acts as a crucial regulator of GABA(A) receptor homeostasis. It may play an important role in the development of CRC and may act as a valuable prognostic indicator of CRC.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70286

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omar Loss et al.
Journal of neuroscience research, 93(7), 1056-1066 (2015-02-06)
Neuronal function requires regulated anterograde and retrograde trafficking of mitochondria along microtubules by using the molecular motors kinesin and dynein. Previous work has established that trafficking kinesin proteins (TRAKs),TRAK1 and TRAK2, are kinesin adaptor proteins that link mitochondria to kinesin
Sai Prasad N Iyer et al.
The Journal of biological chemistry, 278(7), 5399-5409 (2002-11-19)
The abundant and dynamic post-translational modification of nuclear and cytosolic proteins by beta-O-linked N-acetylglucosamine (O-GlcNAc) is catalyzed by O-GlcNAc transferase (OGT). Here we used the yeast two-hybrid approach to identify and isolate GABA(A) receptor-associated protein, GRIF-1 (Beck, M., Brickley, K.
Yanxin An et al.
International journal of colorectal disease, 26(11), 1397-1404 (2011-05-17)
MGb2, a mouse-derived monoclonal antibody specific to gastric carcinoma, was developed in our laboratory. Nevertheless, the potential role of MGb2-antigen/TRAK1 (MGb2-Ag/TRAK1) in colorectal cancer (CRC) is unclear. The aim of this study was to investigate the relationship between MGb2-Ag/TRAK1 expression
Sandra L Gilbert et al.
Nature genetics, 38(2), 245-250 (2005-12-29)
Hypertonia, which results from motor pathway defects in the central nervous system (CNS), is observed in numerous neurological conditions, including cerebral palsy, stroke, spinal cord injury, stiff-person syndrome, spastic paraplegia, dystonia and Parkinson disease. Mice with mutation in the hypertonic
Ortal Barel et al.
Brain : a journal of neurology, 140(3), 568-581 (2017-04-02)
Cellular distribution and dynamics of mitochondria are regulated by several motor proteins and a microtubule network. In neurons, mitochondrial trafficking is crucial because of high energy needs and calcium ion buffering along axons to synapses during neurotransmission. The trafficking kinesin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service