Skip to Content
Merck
All Photos(2)

Documents

HPA005825

Sigma-Aldrich

Anti-SERPINF1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EPC-1 antibody produced in rabbit, Anti-PEDF antibody produced in rabbit, Anti-Pigment epithelium-derived factor precursor antibody produced in rabbit, Anti-Serpin-F1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KVPVNKLAAAVSNFGYDLYRVRSSMSPTTNVLLSPLSVATALSALSLGAEQRTESIIHRALYYDLISSPDIHGTYKELLDTVTAPQKNLKSASRIVFEKKLRIKSSFVAPLEKSYGTRPRVLTGNPRLDLQEINNWVQAQMKGKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINF1(5176)

Immunogen

Pigment epithelium-derived factor precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Pigment epithelium-derived factor (PEDF) is a multifunctional secreted protein encoded by the SERPINF1 gene in humans. It is also referred as OI6, OI12, PEDF, EPC-1 and PIG35. It is more up-regulated in the involuting phase of hemangioma tissues than in the proliferating. It helps in accelerating the involution of hemangioma by growth inhibition of HemECs in an autocrine manner. It may act as a useful marker for evaluating the progression of diabetic nephropathy (DN). Homozygous mutations in this gene causes deficiency of pigment epithelium-derived factor (PEDF) and lead to osteogenesis imperfecta (OI) type VI.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84495

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaine Hui et al.
The Journal of clinical endocrinology and metabolism, 99(11), E2169-E2177 (2014-08-29)
Pigment epithelium-derived factor (PEDF), a circulating glycoprotein with antiangiogenic, antioxidative, and anti-inflammatory properties, protects against diabetic nephropathy (DN) in animal models. We investigated whether circulating PEDF predicted the progression of DN in a 4-year prospective study. Baseline plasma PEDF levels
Hadil Al-Jallad et al.
The Journal of clinical endocrinology and metabolism, 99(11), E2446-E2450 (2014-08-16)
Homozygous mutations in SERPINF1 cause deficiency of pigment epithelium-derived factor (PEDF) and lead to osteogenesis imperfecta (OI) type VI, but it is not known whether heterozygous mutations in SERPINF1 cause a phenotype. In the present study, we therefore assessed family
Kyung-Jin Kim et al.
Biochemical and biophysical research communications, 454(2), 282-288 (2014-12-03)
Hemangioma is a benign tumor derived from abnormal blood vessel growth. Unlike other vascular tumor counterparts, a hemangioma is known to proliferate during its early stage but it is followed by a stage of involution where regression of the tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service