Skip to Content
Merck
All Photos(3)

Key Documents

HPA004869

Sigma-Aldrich

Anti-USP8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HumORF8, Anti-KIAA0055, Anti-SPG59, Anti-UBPY

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.4 μg/mL

immunogen sequence

LPDDSKDTWKKRGNVEYVVLLDWFSSAKDLQIGTTLRSLKDALFKWESKTVLRNEPLVLEGGYENWLLCYPQYTTNAKVTPPPRRQNEEVSISLDFTYPSLEESIPSKPAAQTPPASIEVDENIELISGQNERMGPLNI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... USP8(9101)

Immunogen

Ubiquitin carboxyl-terminal hydrolase 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

USP8 (ubiquitin specific peptidase 8) is a deubiquitinating enzyme. It is involved in the maintenance of endosomal morphology and organization by ubiquitination of the proteins. It is also associated with the cargo trafficking by regulating the chemokine receptor 4 (CXCR4) during endosomal transition. USP8 is also involved in the acrosomogenesis in vivo. It directly interacts with spermatid ESCRT-0 (endosomal-sorting complex required for transport-0) and microtubule structures to form the acrosome. After formation of the acrosomes, USP8 connects acrosome to microtubules via microtubule interacting and trafficking/transport (MIT) domain followed by acrosome assembly and shaping. Mutation in USP8 gene causes Cushing′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86841.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silviu Sbiera et al.
PloS one, 11(12), e0169009-e0169009 (2016-12-23)
Cushing's disease (CD), also known as pituitary-dependent hyperadrenocorticism, is caused by adrenocorticotropic hormone (ACTH)-secreting pituitary tumours. Affected humans and dogs have similar clinical manifestations, however, the incidence of the canine disease is thousand-fold higher. This makes the dog an obvious
Céline Reverdy et al.
Chemistry & biology, 19(4), 467-477 (2012-04-24)
The human USP7 deubiquitinating enzyme was shown to regulate many proteins involved in the cell cycle, as well as tumor suppressors and oncogenes. Thus, USP7 offers a promising, strategic target for cancer therapy. Using biochemical assays and activity-based protein profiling
Isabel Weigand et al.
The Journal of clinical endocrinology and metabolism, 104(7), 2535-2546 (2019-03-08)
Cushing disease (CD) is a rare disorder with severe sequels and incompletely understood pathogenesis. The underlying corticotroph adenomas harbor frequently somatic mutations in the ubiquitin-specific peptidase 8 (USP8) gene. These mutations render USP8 hyperactive and prevent client proteins from degradation.
Zoi Alexopoulou et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(32), E4688-E4697 (2016-07-23)
In Parkinson's disease, misfolded α-synuclein accumulates, often in a ubiquitinated form, in neuronal inclusions termed Lewy bodies. An important outstanding question is whether ubiquitination in Lewy bodies is directly relevant to α-synuclein trafficking or turnover and Parkinson's pathogenesis. By comparative
Giovanna Berruti et al.
Biology of reproduction, 82(5), 930-939 (2010-02-05)
Ubiquitin-specific peptidase 8 (USP8) is a deubiquitinating enzyme that works as a regulator of endosomal sorting and vesicle morphology in cultured cells. Its function in vivo is, however, unknown as USP8 gene deletion leads to embryonic lethality. Previously, we have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service