Skip to Content
Merck
All Photos(2)

Key Documents

HPA004170

Sigma-Aldrich

Anti-CAPN10 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CANP 10 antibody produced in rabbit, Anti-Calcium-activated neutral proteinase 10 antibody produced in rabbit, Anti-Calpain-10 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

FWLPLLEKVYAKVHGSYEHLWAGQVADALVDLTGGLAERWNLKGVAGSGGQQDRPGRWEHRTCRQLLHLKDQCLISCCVLSPRAGARELGEFHAFIVSDL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAPN10(11132)

Related Categories

General description

Calpain 10 (CAPN10), member of the calpain-like cysteine protease family, is an intracellular calcium-dependent non-lysosomal cysteine protease which acts as type 2 diabetes susceptibility gene. It is ubiquitously present in various tissues such as lens, retina, brain, heart, and skeletal muscle. Its large catalytic subunit is composed of domain I (autolytic activation), domain II (cysteine catalytic site), domain III (“electrostatic switch”), and domain IV (calmodulin-like calcium binding sites).

Immunogen

Calpain-10 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Calpain 10 (CAPN10) is majorly associated with the direct regulation of insulin-stimulated glucose uptake in human skeletal muscle cells. It has also reported that, the expression level of CAPN10 has no effects on the insulin-stimulated glycogen synthesis and insulin-signaling pathway via PKB.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86745

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farajollah Maleki et al.
Clinical laboratory, 60(4), 663-670 (2014-05-02)
Calpain-10 is a ubiquitously expressed protease that serves as an intracellular calcium-dependent cysteine protease and is regarded to be one of the candidate genes for type 2 diabetes mellitus (T2DM). We aimed to identify the association of the common variants
Audrey E Brown et al.
Molecular genetics and metabolism, 91(4), 318-324 (2007-06-19)
Calpain-10 was identified as a novel type 2 diabetes susceptibility gene, although the mechanisms by which it increases susceptibility to type 2 diabetes remain unclear. As skeletal muscle is the principal site of the peripheral insulin resistance for glucose disposal
H Ma et al.
The Journal of biological chemistry, 276(30), 28525-28531 (2001-05-29)
Calpains are calcium-dependent intracellular nonlysosomal proteases that are believed to hydrolyze specific substrates important in calcium-regulated signaling pathways. Recently, an atypical member of the calpain family, calpain 10, was described, and genetic variation in this gene was associated with an
Y Horikawa et al.
Nature genetics, 26(2), 163-175 (2000-10-04)
Type 2 or non-insulin-dependent diabetes mellitus (NIDDM) is the most common form of diabetes worldwide, affecting approximately 4% of the world's adult population. It is multifactorial in origin with both genetic and environmental factors contributing to its development. A genome-wide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service