Skip to Content
Merck
All Photos(2)

Documents

HPA003140

Sigma-Aldrich

Anti-TSPAN7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A15, Anti-CD231, Anti-DXS1692E, Anti-MRX58, Anti-MXS1, Anti-TALLA-1, Anti-TM4SF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

TFLRTYTDAMQTYNGNDERSRAVDHVQRSLSCCGVQNYTNWSTSPYFLEHGIPPSCCMNETDCNPQDLHNLTVAATKVNQKGCYDLVTSFMET

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSPAN7(7102)

Immunogen

tetraspanin 7 recombinant protein epitope signature tag (PrEST)

Application

Anti-TSPAN7 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

The TSPAN7 gene encodes a protein known as tetraspanin 7 belonging to the transmembrane 4 superfamily/tetraspanin family. It is involved in filopodia and dendritic spine formation in cultured hippocampal neurons. The protein is essential for the maintenance of spine stability and for normal synaptic transmission. It is involved in the regulation of PICK1 (protein interacting with C kinase 1) and GluA2/3 association, and AMPA receptor trafficking. The morphological and functional maturation of glutamatergic synapses is controlled by this protein.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74399

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerry A McLaughlin et al.
Diabetes, 65(6), 1690-1698 (2016-03-10)
The presence of autoantibodies to multiple-islet autoantigens confers high risk for the development of type 1 diabetes. Four major autoantigens are established (insulin, glutamate decarboxylase, IA2, and zinc transporter-8), but the molecular identity of a fifth, a 38-kDa membrane glycoprotein
Silvia Bassani et al.
Bioarchitecture, 2(3), 95-97 (2012-08-11)
Tetraspanins regulate the signaling, trafficking and biosynthetic processing of associated proteins, and may link the extracellular domain of α-chain integrins with intracellular signaling molecules, including PI4K and PKC, both of which regulate cytoskeletal architecture. We showed that TSPAN7, a member
Daniela Wuttig et al.
International journal of cancer, 131(5), E693-E704 (2012-01-04)
Currently used clinicopathological parameters are insufficient for a reliable prediction of metastatic risk and disease-free survival (DFS) of patients with clear-cell renal cell carcinoma (ccRCC). To identify prognostic genes, the expression profiles of primary ccRCC obtained from patients with different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service