Skip to Content
Merck
All Photos(1)

Key Documents

HPA002844

Sigma-Aldrich

Anti-FBXO18 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-F-box DNA helicase 1 antibody produced in rabbit, Anti-F-box only protein 18 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

HQMTHDGYLKLWQLSKPSLASFDAIFVDEAQDCTPAIMNIVLSQPCGKIFVGDPHQQIYTFRGAVNALFTVPHTHVFYLTQSFRFGVEIAYVGATILDVCKRVRKKTLVGGNHQSGIRGDAK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBXO18(84893)

General description

FBXO18, also known as FBH1 and Fbx18, belongs to the UvrD family of DNA helicases and the superfamily I helicase. It consists of a well conserved F-box motif at its N terminus.

Immunogen

F-box only protein 18 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FBXO18 plays an essential role in the response to DNA replication stress. During DNA replication stress, it promotes double-strand breakage and activation of the DNA-PK and ATM signaling cascades in a helicase-dependent manner. The gene is often deleted or mutated in melanoma cells, which results in their increased survival in response to replicative stress. The gene inactivation may contribute to oncogenic transformation by allowing survival of cells undergoing replicative stress due to external factors (such as UV irradiation). It may be associated with other DNA helicases in tightly controlling cellular homologous recombination (HR) activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86591

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeon-Tae Jeong et al.
Cell cycle (Georgetown, Tex.), 12(7), 1128-1132 (2013-03-08)
FBH1 is a member of the UvrD family of DNA helicases and plays a crucial role in the response to DNA replication stress. In particular, upon DNA replication stress, FBH1 promotes double-strand breakage and activation of the DNA-PK and ATM
Jaehoon Kim et al.
The Journal of biological chemistry, 277(27), 24530-24537 (2002-04-17)
We have identified a novel DNA helicase in humans that belongs to members of the superfamily I helicase and found that it contains a well conserved F-box motif at its N terminus. We have named the enzyme hFBH1 (human F-box
Kasper Fugger et al.
The Journal of cell biology, 186(5), 655-663 (2009-09-09)
Homologous recombination (HR) is essential for faithful repair of DNA lesions yet must be kept in check, as unrestrained HR may compromise genome integrity and lead to premature aging or cancer. To limit unscheduled HR, cells possess DNA helicases capable

Protocols

Subcellular protein location often correlates with function; nucleus proteins implicated in gene regulation, mitochondria proteins in energy production.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service