Skip to Content
Merck
All Photos(7)

Key Documents

HPA002560

Sigma-Aldrich

Anti-SERPINA3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ACT, Anti-Alpha-1-antichymotrypsin precursor, Anti-Cell growth-inhibiting gene 24/25 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

QDKMEEVEAMLLPETLKRWRDSLEFREIGELYLPKFSISRDYNLNDILLQLGIEEAFTSKADLSGITGARNLAVSQVVHKAVLDVFEEGTEASAATAVKITLLSALVETRTIVRFNRPFLMIIVPTDTQNIFFMSKVTNPKQEGAPHR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINA3(12)

General description

SERPINA3 (serpin family A member 3) is a widely expressed protein that belongs to the serpin superfamily. It is synthesized by hepatocytes, bronchial epithelial cells and neuronal cells. It consists of a highly conserved fold and three β-sheets along with ~8 α-helices. In addition to the α, β region, it has a solvent-exposed stretch of amino acids called as the reactive center loop (RCL).

Immunogen

Alpha-1-antichymotrypsin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SERPINA3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SERPINA3 (serpin family A member 3) controls the release of leukocyte proteases during an inflammatory response. It has inhibiting nature for several proteases such as mast cell chymase, pancreatic chymotrypsin, human glandular kallikrein 2, kallikrein 3 (prostate-specific antigen), pancreatic cationic elastase and an uncharacterized lung serum protease. It inhibits the function of neutrophil cathepsin G, a pro-inflammatory enzyme and mast cell chymase, which is responsible for the conversion of angiotensin-1 to the active angiotensin-2. SERPINA3 also restricts inflammation, coagulation and extracellular matrix (ECM) remodeling. Overexpression of the protein is responsible for Alzheimer disease and cerebral amyloid encephalopathy. Deficiency of SERPINA3 causes lung and liver disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST82413

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Rubin et al.
The Journal of biological chemistry, 265(2), 1199-1207 (1990-01-15)
Human alpha 1-antichymotrypsin has been cloned, sequenced and expressed in Escherichia coli and recombinant protein as well as point-specific mutants have been purified and characterized. The corrected gene-deduced amino acid sequence has 45% overall identity with alpha 1-protease inhibitor, which
J P Faber et al.
Journal of hepatology, 18(3), 313-321 (1993-07-01)
Alpha 1-antichymotrypsin (alpha 1-ACT) is a serine proteinase inhibitor (serpin) with cathepsin G, mast cell chymase and chymotrypsin as target enzymes. We present the case of a middle-aged man with low plasma levels of alpha 1-ACT, asthma with progression to
Mónica Santamaria et al.
Gastroenterology, 144(4), 818-828 (2013-01-09)
α1-Antichymotrypsin (α1-ACT), a member of the serpin family (SERPINA3), is an acute-phase protein secreted by hepatocytes in response to cytokines such as oncostatin M. α1-ACT is a protease inhibitor thought to limit tissue damage produced by excessive inflammation-associated proteolysis. However
Sonia T Chelbi et al.
Human molecular genetics, 21(9), 1968-1978 (2012-01-17)
SERPINA3 (Serpin peptidase inhibitor clade A member 3), also known as a1-antichymotrypsin, is a serine protease inhibitor involved in a wide range of biological processes. Recently, it has been shown to be up-regulated in human placental diseases in association with
Michael J Bannon et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(9), 2191-2199 (2014-03-20)
Chronic drug abuse, craving, and relapse are thought to be linked to long-lasting changes in neural gene expression arising through transcriptional and chromatin-related mechanisms. The key contributions of midbrain dopamine (DA)-synthesizing neurons throughout the addiction process provide a compelling rationale

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service