Skip to Content
Merck
All Photos(7)

Documents

HPA002110

Sigma-Aldrich

Anti-PODXL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Podocalyxin-like protein 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LPETMSSSPTAASTTHRYPKTPSPTVAHESNWAKCEDLETQTQSEKQLVLNLTGNTLCAGGASDEKLISLICRAVKATFNPAQDKCGIRLASVPGSQTVVVKEITIHTKLPAKDVYERLKDKWDELKEAGVSDMKLGD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PODXL(5420)

General description

An anti-adhesive transmembrane protein, PODXL (podocalyxin like), belongs to the sialomucin protein family. It is associated with cancer development and aggressiveness. It is expressed in various cells including epithelial cells of kidney glomeruli, vascular endothelia cells, hematopoietic stem cells and platelets.

Immunogen

Podocalyxin-like protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PODXL antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PODXL (podocalyxin like) acts as an anti-adhesion molecule in kidney open filtration pathways. It interacts with the actin-binding protein ezrin in cancer. Using ezrin and the bridging protein NHERF (Na+/H+) exchanger regulatory factor, it connects to the cytoskeleton to regulate cytoskeletal-related functions in tumor progression. In human, PODXL stimulates the cell adherence capacity to immobilized ligands and vascular endothelial cells in presence of integrins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85156

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Marx et al.
Molecular genetics & genomic medicine, 9(5), e1658-e1658 (2021-03-30)
Podocalyxin (PODXL) is a highly sialylated adhesion glycoprotein that plays an important role in podocyte's physiology. Recently, missense and nonsense dominant variants in the PODXL gene have been associated with focal segmental glomerulosclerosis (FSGS), a leading cause of nephrotic syndrome
Steven Sizemore et al.
Cancer research, 67(13), 6183-6191 (2007-07-10)
Podocalyxin is an anti-adhesive transmembrane sialomucin that has been implicated in the development of more aggressive forms of breast and prostate cancer. The mechanism through which podocalyxin increases cancer aggressiveness remains poorly understood but may involve the interaction of podocalyxin
K Boman et al.
British journal of cancer, 108(11), 2321-2328 (2013-05-09)
Membranous expression of the anti-adhesive glycoprotein podocalyxin-like (PODXL) has previously been found to correlate with poor prognosis in several major cancer forms. Here we examined the prognostic impact of PODXL expression in urothelial bladder cancer. Immunohistochemical PODXL expression was examined
Hidenori Kusumoto et al.
Cancer science, 108(3), 528-535 (2016-12-23)
Epithelial-mesenchymal transition (EMT) plays an important role in the progression of lung carcinoma. Podocalyxin (PODXL), which belongs to the CD34 family and regulates cell morphology, has been linked to EMT in lung cancer, and PODXL overexpression is associated with poor
Susana Larrucea et al.
Experimental cell research, 314(10), 2004-2015 (2008-05-06)
Podocalyxin (PODXL) is an anti-adhesive glycoprotein expressed abundantly in the epithelial cells of kidney glomeruli. In contrast, we report herein that expression of podocalyxin(GFP) (PODXL(GFP)) in CHO cells increased the adherence to immobilized fibronectin, spreading, and migration. The transient knockdown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service