Skip to Content
Merck
All Photos(2)

Key Documents

HPA001633

Sigma-Aldrich

Anti-MAP2K7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-JNK kinase 2, Anti-JNK-activating kinase 2, Anti-JNKK 2, Anti-MAP kinase kinase 7, Anti-MAPK/ERK kinase 7, Anti-MAPKK 7, Anti-MP2K7_HUMAN Isoform 2 of O14733 -Homo sapiens, Anti-O14733-2, Anti-c-Jun N-terminal kinase kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LVDSKAKTRSAGCAAYMAPERIDPPDPTKPDYDIRADVWSLGISLVELATGQFPYKNCKTDFEVLTKVLQEEPPLLPGHMGFSGDFQSFVKDCLTKDHRKRPKYNKLLEHSFIKRYETMEV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP2K7(5609)

Immunogen

Dual specificity mitogen-activated protein kinase kinase 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Dual specificity mitogen-activated protein kinase kinase 7 (also known as MAP kinase kinase 7 or MKK7) is an enzyme encoded by the MAP2K7 gene in humans and belongs to mitogen-activated protein kinase kinase family. It can regulate cellular growth, differentiation and apoptosis. Activation of this gene mediates the anti-hepatoma effect of alpinetin. It may serve as a potential agent for the development of hepatoma therapy. It is a direct activator of the mitogen-activated protein kinase family member c-Jun N-terminal kinase (JNK). It activates JNK by phosphorylation of a threonine and tyrosine residue in a Thr-Pro-Tyr motif within kinase subdomain VIII.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86495

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Michael et al.
Biochemical and biophysical research communications, 341(3), 679-683 (2006-01-31)
Mitogen-activated protein kinase kinase 7 (MKK7) is a direct activator of the mitogen-activated protein kinase family member c-Jun N-terminal kinase (JNK). MKK7 activates JNK via phosphorylation of a threonine and tyrosine residue in a Thr-Pro-Tyr motif within kinase subdomain VIII.
Bo Tang et al.
Oncology reports, 27(4), 1090-1096 (2011-12-14)
Alpinetin is a type of novel plant flavonoid derived from Alpinia katsumadai Hayata, found to possess strong anti-hepatoma effects. However, the detailed antitumor mechanism of alpinetin remains unclear. Mitogen-activated protein kinase kinase-7 (MKK7) can regulate cellular growth, differentiation and apoptosis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service