Skip to Content
Merck
All Photos(8)

Key Documents

HPA001222

Sigma-Aldrich

Anti-DGCR14 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DiGeorge syndrome critical region 14 antibody produced in rabbit, Anti-ES2 protein antibody produced in rabbit, Anti-Protein DGCR14 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LRVEGSETPYVDRTPGPAFKILEPGRRERLGLKMANEAAAKNRAKKQEALRRVTENLASLTPKGLSPAMSPALQRLVSRTASKYTDRALRASYTPSPARSTHLKTPASGLQTPTSTPAPGSATRTPLTQDPASITDNL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGCR14(8220)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Protein DGCR14 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DGCR14 (DiGeorge syndrome critical region gene 14) gene is located in the minimal DGS critical region (MDGCR) on chromosome 22q11. This region contains genes responsible for several disorders, such as DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome, and some familial or sporadic conotruncal cardiac defects. The protein forms a component of the C complex spliceosomes and may be involved in pre-mRNA splicing.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73408

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sayuri Takahashi et al.
Scientific reports, 13(1), 12355-12355 (2023-08-01)
Molecular targeted therapy using poly (ADP-ribose) polymerase inhibitors has improved survival in patients with castration-resistant prostate cancer (CRPC). However, this approach is only effective in patients with specific genetic mutations, and additional drug discovery targeting epigenetic modulators is required. Here
W Gong et al.
Human molecular genetics, 5(6), 789-800 (1996-06-01)
The majority of patients with DiGeorge syndrome (DGS) and velo-cardio-facial syndrome (VCFS) have a microdeletion of 22q11. Using translocation breakpoints and fluorescence in situ hybridization analysis (FISH), the minimal DiGeorge critical region (MDGCR) has been narrowed to 250 kb in
Melissa S Jurica et al.
RNA (New York, N.Y.), 8(4), 426-439 (2002-05-07)
We describe characterization of spliceosomes affinity purified under native conditions. These spliceosomes consist largely of C complex containing splicing intermediates. After C complex assembly on an MS2 affinity-tagged pre-mRNA substrate containing a 3' splice site mutation, followed by RNase H

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service