Skip to Content
Merck
All Photos(2)

Key Documents

HPA001084

Sigma-Aldrich

Anti-GPR101 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Probable G-protein coupled receptor 101 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RMEAKDGSLKAKEGSTGTSESSVEARGSEEVRESSTVASDGSMEGKEGSTKVEENSMKADKGRTEVNQCSIDLGEDDMEFGEDDINFSEDDVEAVNIPESLPPSRRNSNSNPPLPRCYQCKAA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR101(83550)

General description

The gene GPR101 (G protein-coupled receptor 101) is mapped to human chromosome Xq26.3 and is mainly expressed in the brain.

Immunogen

Probable G-protein coupled receptor 101 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Probable G-protein coupled receptor 101 is a protein encoded by the GPR101 gene in humans. It modulates cAMP levels and functions in several CNS activities. It is a critical requirement for GnRH-(1-5) transactivation of EGFR (epidermal growth factor receptor) in Ishikawa cells. The gene has only distant identities with other GPCR genes and GPR102 shared identities with GPR57, GPR58 and PNR. It is found to be overexpressed in patients with pituitary lesions. Mutated genes are mostly found in tumors, in patients with acromegaly.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74103

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D K Lee et al.
Gene, 275(1), 83-91 (2001-09-28)
We report the identification, cloning and tissue distributions of ten novel human genes encoding G protein-coupled receptors (GPCRs) GPR78, GPR80, GPR81, GPR82, GPR93, GPR94, GPR95, GPR101, GPR102, GPR103 and a pseudogene, psi GPR79. Each novel orphan GPCR (oGPCR) gene was
Madelaine Cho-Clark et al.
Molecular endocrinology (Baltimore, Md.), 28(1), 80-98 (2013-11-23)
The decapeptide GnRH is known for its central role in the regulation of the hypothalamo-pituitary-gonadal axis. In addition, it is also known to have local effects within peripheral tissues. The zinc metalloendopeptidase, EC 3.4.24.15 (EP24.15), can cleave GnRH at the
Giampaolo Trivellin et al.
The New England journal of medicine, 371(25), 2363-2374 (2014-12-04)
Increased secretion of growth hormone leads to gigantism in children and acromegaly in adults; the genetic causes of gigantism and acromegaly are poorly understood. We performed clinical and genetic studies of samples obtained from 43 patients with gigantism and then
Brian Bates et al.
Brain research, 1087(1), 1-14 (2006-05-02)
This report describes the identification and characterization of the murine orphan GPCR, Gpr101. Both human and murine genes were localized to chromosome X. Similar to its human ortholog, murine Gpr101 mRNA was detected predominantly in the brain within discrete nuclei.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service