Skip to Content
Merck
All Photos(3)

Key Documents

HPA000653

Sigma-Aldrich

Anti-VASH1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Vasohibin-1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GALGMSRREDLMYKPPAFRTLSELVLDFEAAYGRCWHVLKKVKLGQSVSHDPHSVEQIEWKHSVLDVERLGRDDFRKELERHARDMRLKIGKGTGPPSPTKDRKKDVSSPQRAQSSPHRRNSRSERRPSGDKKTSEPKAMPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VASH1(22846)

Immunogen

Vasohibin-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Vasohibin-1 is a protein encoded by the VASH1 gene in humans. It is an angiogenesis inhibitor that is specifically expressed in human vascular endothelial cells in response to vascular endothelial growth factor and fibroblast growth factor-2. It acts as an endothelium-derived negative feedback regulator of angiogenesis. It is induced by fibroblast growth factor 2 (FGF-2) and vascular endothelial growth factor A (VEGF-A). The gene is related to angiogenesis and poor prognosis in several types of cancers. Overexpression of this gene in colorectal carcinoma (CRC) cells increases malignant potential and promotes metastasis. Vasohibin-1 may act as a new biomarker to provide additional prognostic information in patients with colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70356

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keigo Murakami et al.
Human pathology, 45(3), 589-597 (2014-01-22)
Vasohibin 1, an endothelium-derived negative feedback regulator of angiogenesis, is induced by fibroblast growth factor 2 (FGF-2) and vascular endothelial growth factor A (VEGF-A). In this study, we retrospectively evaluated immunoreactivity of FGF-2 and VEGF-A as well as microvessel density
Tao Zhang et al.
Medical oncology (Northwood, London, England), 31(5), 963-963 (2014-04-22)
Vasohibin-1 (VASH1) is a newly discovered angiogenesis inhibitor that is specifically expressed in human vascular endothelial cells in response to vascular endothelial growth factor and fibroblast growth factor-2. This study aimed to evaluate the expression of VASH1 in non-small cell
Yichao Yan et al.
Medical oncology (Northwood, London, England), 31(2), 816-816 (2013-12-25)
Vasohibin-1 has been recently detected in endothelial cells and identified as a negative feedback regulator of angiogenesis induced by VEGF-A. However, the expression of Vasohibin-1 in colorectal cancer and its correlations with VEGF-A, microvessel density (MVD), and the prognosis of
Takahito Kitajima et al.
Anticancer research, 34(10), 5321-5329 (2014-10-03)
Vasohibin-1 (VASH1) is related to angiogenesis and poor prognosis in several types of cancers. However, the biological function and clinical significance of VASH1 in colorectal cancer (CRC) are not fully known. The associations between VASH1 expression and clinicopathological features were
T Kosaka et al.
British journal of cancer, 108(10), 2123-2129 (2013-04-18)
We recently isolated vasohibin-1 (VASH1), a novel angiogenic molecule that is specifically expressed in activated vascular endothelial cells (ECs), and the status of VASH1 expression has been documented in various cancer angiogenesis. The aim of this study was to assess

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service