Skip to Content
Merck
All Photos(7)

Key Documents

HPA000634

Sigma-Aldrich

Anti-IGBP1 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α 4 Protein antibody produced in rabbit, Anti-α4 Protein antibody produced in rabbit, Anti-B cell signal transduction molecule α 4 antibody produced in rabbit, Anti-B cell signal transduction molecule α4 antibody produced in rabbit, Anti-CD79a-binding protein 1 antibody produced in rabbit, Anti-Immunoglobulin-binding protein 1 antibody produced in rabbit, Anti-Renal carcinoma antigen NY-REN-16 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AYPSLVAMASQRQAKIQRYKQKKELEHRLSAMKSAVESGQADDERVREYYLLHLQRWIDISLEEIESIDQEIKILRERDSSREASTSNSSRQERPPVKPFILTRNMAQAKVFGAGYPSLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGBP1(3476)

Looking for similar products? Visit Product Comparison Guide

Immunogen

immunoglobulin binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Immunoglobulin-binding protein 1 (also known as IBP1, α-4) is a protein encoded by the IGBP1 gene in humans and is located at q13.1-q13.3 on chromosome X. It is a regulatory subunit of the major cellular phosphatase. An altered expression of this gene through either a change in translational efficiency, mRNA stability or splicing may be associated with Opitz GBBB syndrome. The gene is associated with a variety of functions that regulate many cellular processes and also possesses an oncogenic function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74044

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John M Graham et al.
American journal of medical genetics. Part A, 123A(1), 37-44 (2003-10-14)
We describe two brothers with a unique pattern of malformations that includes coloboma (iris, optic nerve), high forehead, severe retrognathia, mental retardation, and agenesis of the corpus callosum (ACC). Both boys have low-set cupped ears with sensorineural hearing loss, normal
L-P Chen et al.
Oncogene, 30(26), 2943-2953 (2011-02-23)
A regulator of the protein phosphatase 2A (PP2A), α4, has been implicated in a variety of functions that regulate many cellular processes. To explore the role of α4 in human cell transformation and tumorigenesis, we show that α4 is highly
M Onda et al.
Genomics, 46(3), 373-378 (1998-01-27)
To study the function of the B cell signal transduction molecule alpha 4 (IGBP1), we isolated a human alpha 4 (IGBP1) gene that has sequence similarity to the yeast protein (TAP42) involved in the rapamycin-sensitive signal transduction pathway. The human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service