product line
MISSION®
form
solid
mature sequence
AAAGUGCAUCUUUUUAGAGGAU
Sanger mature/minor accession no.
Sanger microRNA accession no.
storage temp.
−20°C
Gene Information
General description
- Optimized and ready for transfection.
- Novel MISSION miRNA mimic design has been functionally tested for knockdown efficiency against natural miRNA targets.
- Unique MISSION miRNA mimic design significantly reduces possible sense strand off target effects.
- Available as a whole human library and individual miRNA targets.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service