Select a Size
$462.00
Estimated to ship onMay 31, 2025
Select a Size
About This Item
$462.00
Estimated to ship onMay 31, 2025
Recommended Products
shipped in
ambient
storage temp.
room temp
General description
Application
- Optical clearing agent in tissue imaging studies
- Refractive index matching agent for microscopy
Visikol HISTO products are easy-to-use and ideal for rapid tissue clearing applications. Tissue clearing is completely reversible with no alteration to the tissue.
Features and Benefits
- Render tissue samples transparent for imaging.
- Achieve clearer, sharper images in antibody tissue staining applications.
- The Visikol HISTO tissue clearing process does not affect prior immunolabeling.
- Save time. The Visikol HISTO method is rapid relative to many popular current tissue clearing techniques.
- Retain morphology and lipid content in your samples.
- The HISTO tissue clearing method is easily reversible for follow up 2D histology/IHC or for 3D data validation against traditional histology.
- Non-toxic.
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3
Target Organs
Eyes,Central nervous system, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service