Skip to Content
Merck
All Photos(1)

Documents

H9037

Sigma-Aldrich

Anti-HA−TRITC antibody, Mouse monoclonal

~1 mg/mL, clone HA-7, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

conjugate

TRITC conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HA-7, monoclonal

form

buffered aqueous solution

storage condition

protect from light

concentration

~1 mg/mL

technique(s)

direct immunofluorescence: 10-15 mg/mL using mammalian cells expressing HA-tagged fusion proteins

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Monoclonal Anti-HA is derived from the HA-7 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Monoclonal anti-HA recognizes the HA tag sequence on HA tagged fusion proteins by immunocytochemistry. The antibody reacts specifically with N- and C-terminal HA tagged fusion proteins.

Specificity

The antibody recognizes native as well as denatured-reduced forms of HA-tagged proteins and is reactive with N- or C-terminal HA-tagged fusion proteins expressed in E. coli or in mammalian cells.

Immunogen

synthetic peptide corresponding to a fragment of human influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Monoclonal Anti-HA-TRITC antibody was used to test if DOT/ICM effector SDHA is necessary for virulence of Legionella pneumophila in Galleria mellonella and A/J mice.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 1% bovine serum albumin 15 mM sodium azide as a preservative.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Yamazaki et al.
Science immunology, 5(44) (2020-03-01)
We investigated the molecular and cellular basis of severe combined immunodeficiency (SCID) in six patients with otofaciocervical syndrome type 2 who failed to attain T cell reconstitution after allogeneic hematopoietic stem cell transplantation, despite successful engraftment in three of them.
D J Scott et al.
British journal of urology, 58(2), 119-124 (1986-04-01)
To assess the effectiveness of the surgical correction of vesico-ureteric reflux, current renal function was determined in 56 children with scarred kidneys who had undergone ureteric reimplantation between 1978 and 1983. The children were aged between 2 and 15 years
Clare R Harding et al.
Infection and immunity, 81(7), 2598-2605 (2013-05-08)
Legionella pneumophila is an intracellular bacterium that resides within amoebae and macrophages in a specialized compartment termed the Legionella-containing vacuole (LCV). As well as providing an intracellular niche for replication, the LCV helps to prevent the release of bacterial components
P O Olins et al.
Current opinion in biotechnology, 4(5), 520-525 (1993-10-01)
Recent advances in protein expression in E. coli have focused primarily on the enhancement of protein quality. Problems in mRNA translation such as inefficient initiation, mistranslation, frame-shifting and frame-hopping can often be addressed by altering heterologous gene-coding sequences. Fusion technology
Gunnar N Schroeder et al.
Infection and immunity, 83(10), 3989-4002 (2015-07-29)
Legionella pneumophila is a bacterial pathogen that thrives in alveolar macrophages, causing a severe pneumonia. The virulence of L. pneumophila depends on its Dot/Icm type IV secretion system (T4SS), which delivers more than 300 effector proteins into the host, where

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service