Skip to Content
Merck
All Photos(1)

Key Documents

H7877

Sigma-Aldrich

Serotonin hydrogenoxalate

powder

Synonym(s):

3-(2-Aminoethyl)-5-hydroxyindole hydrogenoxalate, 5-HT, 5-Hydroxytryptamine hydrogenoxalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O · C2H2O4
CAS Number:
Molecular Weight:
266.25
Beilstein:
3792887
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

powder

storage temp.

2-8°C

SMILES string

OC(=O)C(O)=O.NCCc1c[nH]c2ccc(O)cc12

InChI

1S/C10H12N2O.C2H2O4/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;3-1(4)2(5)6/h1-2,5-6,12-13H,3-4,11H2;(H,3,4)(H,5,6)

InChI key

VTBOICJSERLVPD-UHFFFAOYSA-N

Gene Information

human ... HTR3A(3359)

Looking for similar products? Visit Product Comparison Guide

Application

Serotonin hydrogenoxalate has been used to study serotonin-mediated release of catecholamine from the chromaffin tissue of the atlantic hagfish (Myxine glutinosa L.).[1]

Biochem/physiol Actions

Neurotransmitter.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernier et al.
The Journal of experimental biology, 199(Pt 11), 2485-2497 (1996-01-01)
An in situ saline-perfused systemic heart/posterior cardinal vein preparation of the Atlantic hagfish (Myxine glutinosa) was used to assess (1) the ability of the chromaffin tissue to release catecholamines in response to adrenocorticotropic hormone (ACTH; 7.5 i.u. kg-1), serotonin (250
Hitoshi Aonuma et al.
Neurobiology of learning and memory, 141, 189-198 (2017-04-30)
The pond snail Lymnaea stagnalis is capable of learning taste aversion by pairing presentations of a sucrose solution and an electric shock and consolidating it into long-term memory (LTM), which is referred to as conditioned taste aversion (CTA). We asked
Wen-Wen Liu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(5), 6570-6581 (2020-04-05)
Dysfunction of the circadian rhythm is one of most common nonmotor symptoms in Parkinson's disease (PD), but the molecular role of the circadian rhythm in PD is unclear. We here showed that inactivation of brain and muscle ARNT-like 1 (BMAL1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service