Skip to Content
Merck
All Photos(1)

Key Documents

H6785

Sigma-Aldrich

15(S)-Hydroxy-(5Z,8Z,11Z,13E)-eicosatetraene-(2-biotinyl)hydrazide

≥98%, ethanol solution

Synonym(s):

15(S)-HETE−biotin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48N4O4S
CAS Number:
Molecular Weight:
560.79
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

ethanol solution

εmax

23,000 at 236 nm

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCC[C@H](O)\C=C\C=C/C\C=C/C\C=C/CCCC(=O)NNC(=O)CCCC[C@@H]1SCC2NC(=O)NC12

InChI

1S/C30H48N4O4S/c1-2-3-13-18-24(35)19-14-11-9-7-5-4-6-8-10-12-15-21-27(36)33-34-28(37)22-17-16-20-26-29-25(23-39-26)31-30(38)32-29/h4-5,8-11,14,19,24-26,29,35H,2-3,6-7,12-13,15-18,20-23H2,1H3,(H,33,36)(H,34,37)(H2,31,32,38)/b5-4-,10-8-,11-9-,19-14+/t24-,25-,26-,29-/m0/s1

InChI key

CVMXPPQBNJUHBS-DJDVYCAUSA-N

Application

May be used to detect 15(S)-HETE binding proteins and/or receptors.

Biochem/physiol Actions

Potent 5-lipoxygenase inhibitor.

Other Notes

15(S)-HETE is a major arachidonic acid metabolite produced from the 15-lipoxygenase pathway.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service